Skip to Content
Merck
All Photos(1)

Key Documents

SLLGR04NL

Millipore

Millex hydrophilic PTFE syringe filter

pore size 0.2 μm, diam. 4 mm, non-sterile

Synonym(s):

0.2 μm PTFE syringe filter, Millex-LG syringe filter, disposable syringe filter, syringe filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
eCl@ss:
32031690
NACRES:
NB.22

material

PTFE membrane
high-density polyethylene housing

sterility

non-sterile

product line

Millex

feature

holdup volume< 10 μL
hydrophilic

manufacturer/tradename

Millex

parameter

14 bar max. inlet pressure (200 psi)
45 °C max. temp.

technique(s)

LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable
analytical sample preparation: suitable
mass spectrometry (MS): suitable

H

19.8 mm

diam.

4 mm

filtration area

0.1 cm2

volume

1 mL

pore size

0.2 μm

input

sample type aqueous solution(s)
sample type organic solution(s)

fitting

female Luer-Lok® inlet
male Luer outlet stepped slip

shipped in

ambient

General description

Filter Code: LG
Overview of Non-Sterile Millex Syringe Filters with Hydrophilic PTFE
• Lowest extractables, optimizing background levels of sensitive analyses like UHPLC and LC-MS
• Filtration of protein-containing solutions, and aqueous or organic solutions
• Available in 0.20 µm and 0.45 µm pore sizes and three diameters to suit your application needs

Available in Multiple Pack Sizes:
SKU ending with NS (-------NS): 50 per pack
SKU ending with NL (-------NL): 100 per pack
SKU ending with NB (-------NB): 250 per pack
SKU ending with NK (-------NK): 1000 per pack
SLCRT13NL: 100 per pack. These syringe filters have a tube outlet

Applications:
Sample Filtration Prior to UHPLC, HPLC and Mass Spec; Solvent Filtration; Clarification of Aqueous and Organic Solutions

Application

0.20 µm pore size, 4 mm diameter, Millex-LG hydrophilic PTFE membrane

Packaging

non sterile

Specifications

For general laboratory use only

Physical form

Color Code: Natural inlet/natural outlet

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
Millex is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Fan et al.
mAbs, 12(1), 1742457-1742457 (2020-03-28)
Ebola virus (EBOV) can cause severe hemorrhagic fever in humans, and no approved treatment is currently available. Although several antibodies have achieved good protection in animal models, the potential emerging isolates of ebolavirus and the unknown effects of experimental antibodies

Articles

Prior to probing a membrane using precious antibodies, it is helpful to visualize the transferred proteins on the blot to ensure complete transfer and even loading. This page describes possible causes and potential remedies for challenges encountered during protein visualization.

The possible causes and potential remedies for challenges encountered as a result of blocking, washing, antibody incubation, and detection/exposure of Western blots

Millex® hydrophilic PTFE syringe filters offer broad chemical compatibility and robust performance for various sample filtrations.

Millex® hydrophilic PTFE syringe filters offer broad chemical compatibility and robust performance for various sample filtrations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service