Skip to Content
Merck
All Photos(1)

Key Documents

H-004

Supelco

Hydromorphone solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@@]23[C@H]4CCC(=O)[C@@H]2Oc5c(O)ccc(C[C@@H]14)c35

InChI

1S/C17H19NO3/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18/h2,4,10-11,16,19H,3,5-8H2,1H3/t10-,11+,16-,17-/m0/s1

InChI key

WVLOADHCBXTIJK-YNHQPCIGSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

A certified reference standard suitable for use as a starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research. Hydromorphone, marketed under brand names including Palladone and Dilaudid®, is a very potent opioid analgesic. The drug is often used medicinally as an alternative to morphine for treatment of pain or as a cough suppressant for painful coughing resulting from continued bronchial irritation.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dilaudid is a registered trademark of Purdue Pharma L.P.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandon A Van Noord et al.
Journal of clinical anesthesia, 25(2), 146-149 (2013-01-22)
A 35 year old woman, 6 days after ileal neobladder construction, reported uncontrolled pain despite 33 mg hydromorphone via patient-controlled analgesia (PCA). Abdominal compartment syndrome was suspected based on worsening tachypnea, oxygen desaturation, and severe, prolonged ileus. Following emergent intubation
Alec B O'Connor et al.
Journal of opioid management, 8(6), 403-413 (2012-12-25)
The reasons providers choose one parenteral opioid over another are not well understood. The authors sought to determine why emergency department (ED) providers choose one parenteral opioid over another. In a cohort of ED patients who received intravenous morphine or
Martin E Hale et al.
Journal of opioid management, 8(5), 299-314 (2012-12-19)
This analysis was designed to assess the pooled safety and tolerability of once-daily hydromorphone extended release (ER) (OROS® hydromorphone ER) in opioid-naïve and opioid-tolerant patients with chronic cancer or noncancer pain. Safety results were pooled from 13 controlled and uncontrolled
Erik Rauch
AANA journal, 80(4 Suppl), S25-S32 (2012-12-20)
Despite compelling evidence for the safety and efficacy of intrathecal hydromorphone, the use of this opioid intrathecally for the pain management of patients undergoing cesarean delivery has not been widely accepted. The purpose of this retrospective study was to compare
Kyung Hee Lee et al.
Journal of opioid management, 8(4), 243-252 (2012-09-04)
To evaluate the effectiveness of OROS® hydromorphone in reducing breakthrough pain (BTP) medication frequency in Korean patients with chronic cancer pain. Multicenter, prospective, open-label, phase IV study. Patients with chronic malignant pain using immediate-release oxycodone more than two times per

Articles

Although both biphenyl and phenyl-hexyl phases can resolve these compounds, the former exhibits excellent peak shape and substantially less silanol-derived ion exchange activity.

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service