Skip to Content
Merck
All Photos(1)

Key Documents

O-002

Supelco

Oxycodone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO4
CAS Number:
Molecular Weight:
315.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

COc1ccc2C[C@H]3N(C)CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O

InChI

1S/C18H21NO4/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10/h3-4,13,16,21H,5-9H2,1-2H3/t13-,16+,17+,18-/m1/s1

InChI key

BRUQQQPBMZOVGD-XFKAJCMBSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Spiking Solution® is suitable for numerous applications in GC/MS or LC/MS testing of oxycodone from clinical toxicology and urine drug testing to forensic analysis. Oxycodone is a semi-synthetic opioid analgesic sold under many trade names including OxyContin® and Percocet® for treatment of moderate to severe pain. Oxycodone is also sold illicitly under street names such as "Roxies," "Blues," and "Perks".

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
OxyContin is a registered trademark of Purdue Pharma L.P.
Percocet is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Merchant et al.
Journal of opioid management, 9(1), 51-61 (2013-05-28)
To evaluate a composite measure for chronic pain that balances pain relief with tolerability. Post hoc meta-analysis of three randomized, multicenter, double-blind studies. Subjects with moderate-to-severe chronic osteoarthritis knee pain or low back pain who had been randomized to receive
FDA ban of generic oxycodone calls into question Health Canada's approval.
Adam Miller
CMAJ : Canadian Medical Association journal = journal de l'Association medicale canadienne, 185(9), E381-E382 (2013-05-01)
Myoung Kim et al.
Journal of opioid management, 9(3), 189-204 (2013-06-19)
This article describes the rationale and design of the Oxycodone Users Registry (OUR) study and lessons learned during study development and data collection. The OUR study used a prospective registry design. Sixty-five academic and private medical practices across the United
Kenneth Kirsh et al.
Journal of pain & palliative care pharmacotherapy, 26(4), 348-361 (2013-05-16)
Prescription opioids are prescribed increasingly for the management of chronic pain, and this has been accompanied by a dramatic rise in opioid-related abuse, addiction, and overdose deaths. Reports of abuse involving nonoral administration (e.g., snorting, injecting) of prescription opioids are
Reply to commentary.
Stephen F Butler
The journal of pain : official journal of the American Pain Society, 14(4), 361-362 (2013-04-04)

Articles

Although both biphenyl and phenyl-hexyl phases can resolve these compounds, the former exhibits excellent peak shape and substantially less silanol-derived ion exchange activity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service