610015
Avanti
Syringe (250 μL)
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder Syringe, Gas-tight Syringe, 250μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
General description
Replacement Part for Avanti′s Mini-Extruder, products 610000 and 610023. Contains plunger, needle, syringe screw cap, and PTFE seals.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Application
Syringe or Gas-tight Syringe (250 μL) has been used in liposome preparation.
Packaging
Syringe Box (8-3/8" X 1-5/8" X 1-1/4") (610015-1Ea)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
also commonly purchased with this product
Product No.
Description
Pricing
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
S H Li et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 18(4), 1261-1269 (1998-03-14)
Huntingtin is the protein product of the gene for Huntington's disease (HD) and carries a polyglutamine repeat that is expanded in HD (>36 units). Huntingtin-associated protein (HAP1) is a neuronal protein and binds to huntingtin in association with the polyglutamine
Byoungjae Kong et al.
Nature communications, 10(1), 185-185 (2019-01-16)
Membrane-disrupting agents that selectively target virus versus host membranes could potentially inhibit a broad-spectrum of enveloped viruses, but currently such antivirals are lacking. Here, we develop a nanodisc incorporated with a decoy virus receptor that inhibits virus infection. Mechanistically, nanodiscs
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service