610019
Avanti
Extruder Washer-PTFE
Avanti Research™ - A Croda Brand
Synonym(s):
Extruder Washer-PTFE, PTFE Washer for Extruder (1)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151503
NACRES:
NB.51
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
15-25°C
General description
Replacement Part for Avanti′s Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Watch the video to learn more about the Avanti Research™ Mini-Extruder.
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
also commonly purchased with this product
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service