Key Documents
20997
Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
1001TLL, PTFE Luer lock, volume 1 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Pricing and availability is not currently available.
material
PTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81320
needle L
, n/a
volume
1 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1001TLL (PTFE Luer lock)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.[1] The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.[2]
Application
It was used in alkene termination procedure during olefin cross-metathesisIt was used in alkene termination procedure during olefin cross-metathesis to functionalize the surface of gallium nitride (0001) with a carboxyl group for peptide binding.[3]t
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Guy, Jeffrey S.
Pharmacology for the Prehospital Professional, 54-54 (2009)
Covalent attachment of a peptide to the surface of gallium nitride.
Makowski, Matthew S., et al.
Surface Science, 605.15, 1466-1475 (2011)
Questions
1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?
    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
      Helpful?
  2. How can I determine the shelf life / expiration / retest date of this product?
    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
      Helpful?