Skip to Content
Merck
All Photos(1)

Key Documents

W346306

Sigma-Aldrich

4-Methylpentanoic acid

≥98%, FCC, FG

Synonym(s):

4-Methylvaleric acid, 4-Methylpentanoic acid, Isocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2CO2H
CAS Number:
Molecular Weight:
116.16
FEMA Number:
3463
Beilstein:
1741912
EC Number:
Council of Europe no.:
10150
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.057
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥98%

refractive index

n20/D 1.414 (lit.)

bp

199-201 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

pungent; cheesy

SMILES string

CC(C)CCC(O)=O

InChI

1S/C6H12O2/c1-5(2)3-4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)

InChI key

FGKJLKRYENPLQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G E Mortimore et al.
The Journal of biological chemistry, 262(34), 16322-16327 (1987-12-05)
Previous studies with livers from fed rats perfused in the single-pass mode have shown that regulatory amino acids (Leu, Tyr, Gln, Pro, Met, His, and Trp) as a group as well as leucine alone inhibit deprivation-induced protein degradation optimally at
Viridiana Tejada-Ortigoza et al.
Molecules (Basel, Switzerland), 24(4) (2019-02-17)
Fruit by-products are being investigated as non-conventional alternative sources of dietary fiber (DF). High hydrostatic pressure (HHP) treatments have been used to modify DF content as well as its technological and physiological functionality. Orange, mango and prickly pear peels untreated
S C Ling et al.
Journal of applied microbiology, 111(3), 559-571 (2011-06-22)
Polyhydroxyalkanoate (PHA) with enhanced physicochemical properties will be ideal for a wide range of practical applications. The incorporation of 3-hydroxy-4-methylvalerate (3H4MV) into the polymer backbone is known to improve the overall properties of the resulting polymer. However, the most suitable
R A Bowman et al.
Journal of clinical pathology, 39(2), 212-214 (1986-02-01)
A total of 329 selective enrichment broth cultures were tested for detection of Clostridium difficile by latex particle agglutination (LPA), gas-liquid chromatography, and bacterial culture. Of 53 broths positive by LPA, 36 were positive by gas-liquid chromatography, and 42 were
M R Charlton et al.
Diabetes, 46(8), 1336-1340 (1997-08-01)
Although insulin's anticatabolic effect on protein metabolism in type 1 diabetes has been clearly shown to be related to the inhibition of protein breakdown, insulin's effect on muscle protein synthesis remains controversial. Cross-limb studies and measurements of synthesis rates of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service