Skip to Content
Merck
All Photos(1)

Key Documents

W270601

Sigma-Aldrich

2-Methylheptanoic acid

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(CH3)CO2H
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2706
EC Number:
Council of Europe no.:
2003
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.047
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

Assay

≥97%

refractive index

n20/D 1.425 (lit.)

bp

140 °C/30 mmHg (lit.)

density

0.906 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; cheesy; waxy

SMILES string

CCCCCC(C)C(O)=O

InChI

1S/C8H16O2/c1-3-4-5-6-7(2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

NKBWMBRPILTCRD-UHFFFAOYSA-N

General description

2-Methylheptanoic acid has been identified in honey.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ngan N T Nguyen et al.
Scientific reports, 8(1), 15486-15486 (2018-10-21)
Hepatitis C virus (HCV) exploits an extensive network of host proteins to maintain chronic infection. Using RNA-Seq technology, we identified 30 host genes that were differentially expressed in cell culture grown HCV (HCVcc)-infected cells. Of these candidate genes, we selected
Floral markers in honey of various botanical and geographic origins: a review.
Kaskonien? V & Venskutonis PR.
Comprehensive Reviews in Food Science and Food Safety, 9(6), 620-634 (2010)
Paul E Minkler et al.
Molecular genetics and metabolism, 120(4), 363-369 (2017-02-14)
While selectively quantifying acylcarnitines in thousands of patient samples using UHPLC-MS/MS, we have occasionally observed unidentified branched-chain C8 acylcarnitines. Such observations are not possible using tandem MS methods, which generate pseudo-quantitative acylcarnitine "profiles". Since these "profiles" select for mass alone
Gang-Tian Zhu et al.
Journal of chromatography. A, 1564, 42-50 (2018-06-11)
Tailor-made chitosan fiber was prepared via hydrothermal treatment to serve as a micro-solid phase extraction (micro-SPE) sorbent for the analysis of petroleum acids (PAs) in crude oils. Chitosan fiber, which is commercial and cheap, has a diameter of about 10 μm
Andrew T Schuster et al.
Gastroenterology, 148(7), 1405-1416 (2015-02-24)
Defects in colonic epithelial barrier defenses are associated with ulcerative colitis (UC). The proteins that regulate bacterial clearance in the colonic epithelium have not been completely identified. The Drosophila chromosome-associated protein D3 (dCAP-D3) regulates responses to bacterial infection. We examined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service