Skip to Content
Merck
All Photos(1)

Key Documents

W248908

Sigma-Aldrich

Furfural

≥98%, FCC, FG

Synonym(s):

2-Furaldehyde, Furan-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
FEMA Number:
2489
Beilstein:
105755
EC Number:
Council of Europe no.:
2014
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.018
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 175.105

vapor density

3.31 (vs air)

vapor pressure

13.5 mmHg ( 55 °C)

Assay

≥98%

form

liquid

autoignition temp.

599 °F

expl. lim.

19.3 %

refractive index

n20/D 1.525 (lit.)

bp

162 °C (lit.)

mp

−36 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

almond; baked; bread; woody; sweet

SMILES string

[H]C(=O)c1ccco1

InChI

1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H

InChI key

HYBBIBNJHNGZAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Intermolecular Diels-Alder Cycloadditions of Furfural-Based Chemicals from Renewable Resources.: Focuses on the regio- and diastereoselectivity of furfural in reactions with alkenes, highlighting its potential in developing bio-based polymers (Galkin and Ananikov, 2021).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lilong Zhou et al.
Bioresource technology, 129, 450-455 (2012-12-26)
A new kind of bifunctional ionic liquid catalysts was synthesized to degrade microcrystalline cellulose in [BMIM]Cl at atmospheric pressure. The effects of reaction temperature, amount of catalysts, reaction time, ionic liquid purity and cellulose concentration on conversion were investigated. At
Arvind H Jadhav et al.
Bioresource technology, 132, 342-350 (2013-02-26)
Acidity modified silver exchanged silicotungstic acid (AgSTA) catalyst was prepared and characterized by X-ray diffraction, FT-IR spectroscopy, Raman spectroscopy, FT-IR pyridine adsorption, SEM imaging, EDX mapping, and antimicrobial activity was also tested. The catalytic activity was evaluated for the dehydration
Jianghua He et al.
ChemSusChem, 6(1), 61-64 (2012-12-12)
It's nano: Small and uniform chromium nanoparticles, either preformed or generated in situ, effectively catalyze the conversion of glucose into 5-hydroxymethyl furfural. The results compare favorably with those achieved by using a catalyst system based on divalent CrCl(2) in ionic
Reetta Karinen et al.
ChemSusChem, 4(8), 1002-1016 (2011-07-06)
Furfurals are important intermediates in the chemical industry. They are typically produced by homogeneous catalysis in aqueous solutions. However, heterogeneously catalyzed processes would be beneficial in view of the principles of green chemistry: the elimination of homogeneous mineral acids makes
Tim Ståhlberg et al.
ChemSusChem, 4(4), 451-458 (2011-01-29)
The synthesis of 5-(hydroxymethyl)furfural (HMF) in ionic liquids is a field that has grown rapidly in recent years. Unique dissolving properties for crude biomass in combination with a high selectivity for HMF formation from hexose sugars make ionic liquids attractive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service