Skip to Content
Merck
All Photos(1)

Key Documents

W244406

Sigma-Aldrich

Ethyl 3-methyl-3-phenylglycidate

≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
FEMA Number:
2444
EC Number:
Council of Europe no.:
6002
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.015
NACRES:
NA.21
Organoleptic:
strawberry; sweet
grade:
FG
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

Assay

≥98%

refractive index

n20/D 1.505 (lit.)

bp

272-275 °C (lit.)

density

1.087 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

strawberry; sweet

storage temp.

2-8°C

SMILES string

CCOC(=O)C1OC1(C)c2ccccc2

InChI

1S/C12H14O3/c1-3-14-11(13)10-12(2,15-10)9-7-5-4-6-8-9/h4-8,10H,3H2,1-2H3

InChI key

LQKRYVGRPXFFAV-UHFFFAOYSA-N

General description

Ethyl 3-methyl-3-phenylglycidate is commonly used in low-fat strawberry flavored ice-creams due to its strawberry-like flavor.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

267.8 °F - closed cup

Flash Point(C)

131 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of fat level on the perception of five flavor chemicals in ice cream with or without fat mimetics by using a descriptive test.
Liou BK and Grun IU
Journal of Food Science, 72(8), S595?S604-S595?S604 (2007)
Long-term toxicity study of ethyl methylphenylglycidate (strawberry aldehyde) in the rat.
D Dunnington et al.
Food and cosmetics toxicology, 19(6), 691-699 (1981-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service