Skip to Content
Merck
All Photos(1)

Key Documents

W263508

Sigma-Aldrich

Linalool

≥97%, FCC, FG

Synonym(s):

(±)-3,7-Dimethyl-1,6-octadien-3-ol, (±)-3,7-Dimethyl-3-hydroxy-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2635
Beilstein:
1721488
EC Number:
Council of Europe no.:
61
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.013
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor pressure

0.17 mmHg ( 25 °C)

Assay

≥97%

composition

contains EU 1223/2009 restricted linalool

refractive index

n20/D 1.462 (lit.)

bp

194-197 °C/720 mmHg (lit.)

solubility

ethanol: soluble 1ml/4ml, clear, colorless (60% ethanol)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

linalool

Organoleptic

lemon; orange; floral; sweet

SMILES string

C\C(C)=C\CCC(C)(O)C=C

InChI

1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3

InChI key

CDOSHBSSFJOMGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Linalool is a monoterepene compound. It is the major component of many essential oils. Anti-inflammatory properties of (−) linalool, a naturally occurring enantiomer, has been studied. It is also the major constituent of the basil and thyme essential oil. Linalool is reported to have a lemon like odor.
Linalool or ((±)-3,7-Dimethyl-1,6-octadien-3-ol is a reported fragrance ingredient.

Application

Linalool has been used to study the blends of various flower aroma compounds as a floral attractant for the migratory moth velvetbean caterpillar (Anticarsia gemmatalis Hübner).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

171.0 °F - Pensky-Martens closed cup

Flash Point(C)

77.2 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binary floral lure attractive to velvetbean caterpillar adults (Lepidoptera: Noctuidae).
Meagher RL and Landolt PJ.
Florida entomologist, 93(1), 73-79 (2010)
Transformations of monoterpenoids in aqueous acids: The reactions of linalool. geraniol, nerol and their acetates in aqueous citric acid.
Baxter RL ,et al.
Tetrahedron, 34(14), 2195-2199 (1978)
Inhibitory effect of thyme and basil essential oils, carvacrol, thymol, estragol, linalool and p-cymene towards Shigella sonnei and S. flexneri.
Bagamboula CF, et al.
Food Microbiology, 21(1), 33-42 (2004)
H M Schellinck et al.
Chemical senses, 26(6), 663-672 (2001-07-28)
The present paper describes a quick and efficient method for assessing olfactory discrimination learning in mice. In training mice received trials in which one odor (CS+) was paired with sugar and another odor (CS-) was paired with no sugar. When
L Re et al.
Pharmacological research, 42(2), 177-182 (2000-07-11)
Linalool is a monoterpene compound reported to be a major component of essential oils in various aromatic species. Several linalool-producing species are used in traditional medical systems. Among these is Aeolanthus suaveolens G. Dom (Labiatae) which is used as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service