Skip to Content
Merck
All Photos(2)

Key Documents

M54058

Sigma-Aldrich

Methylmalonic acid

99%

Synonym(s):

2-Methylpropanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CO2H)2
CAS Number:
Molecular Weight:
118.09
Beilstein:
1756084
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

128-130 °C (lit.)

SMILES string

CC(C(O)=O)C(O)=O

InChI

1S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)

InChI key

ZIYVHBGGAOATLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Methylmalonic acid can be used as a substrate in:
  • Esterification reactions by various catalysts.
  • Synthesis of pyrimidinium betaines with formamidine.
  • Synthesis of malonic acid half thioester (MAHT).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wendy Schijns et al.
The American journal of clinical nutrition, 108(1), 6-12 (2018-06-23)
After Roux-en-Y gastric bypass (RYGB), patients often develop a vitamin B-12 deficiency. Our objective was to investigate whether oral supplementation increases and normalizes low vitamin B-12 concentrations (vitamin B-12 > 200 pmol/L) in RYGB patients as compared to intramuscular injections. A randomized
Georg Staubli et al.
Pediatric emergency care, 32(12), 827-829 (2015-11-17)
The efficiency of nitrous oxide in an equimolar mixture with oxygen or in concentrations up to 70% is approved for short painful procedures. Evaluation of the vitamin B12 levels in anesthetic staff applying nitrous oxide showed reduced vitamin B12 plasma
A Stable anionic N-heterocyclic carbene and its zwitterionic complexes.
Cesar V, et al.
Journal of the American Chemical Society, 130(34), 11286-11287 (2008)
Novel Bronsted acidic ionic liquid as efficient and reusable catalyst system for esterification.
Gui J, et al.
Catalysis Communications, 5(9), 473-477 (2004)
L S Kornerup et al.
Clinical nutrition (Edinburgh, Scotland), 38(2), 906-911 (2018-03-07)
Bariatric surgery increases the risk of micronutrient deficiencies, including vitamin B12 (B12) deficiency. We analysed early changes in biomarkers of B12 status following bariatric surgery. We prospectively included adult patients (n = 27) referred for either Roux-en-Y Gastric Bypass (RYGB) (n = 19) or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service