Skip to Content
Merck
All Photos(1)

Key Documents

M-105

Supelco

Methyl-D3-malonic acid solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3O4D3
CAS Number:
Molecular Weight:
121.11
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C(C(O)=O)C(O)=O

InChI

1S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)/i1D3

InChI key

ZIYVHBGGAOATLY-FIBGUPNXSA-N

General description

Methylmalonic acid, a malonic acid derivative, is a vital intermediate in the metabolism of fat and proteins and plays a key role in energy metabolism. Elevated methylmalonic acid levels can be caused by an inborn error of metabolism leading to methylmalonic aciduria or indicate early vitamin B<sub>12</sub> deficiency. This internal standard is suitable for quantitation of methylmalonic acid levels by LC/MS or GC/MS in newborn screening and clinical testing applications to test for methylmalonic acidemia and diagnose vitamin B<sub>12</sub> deficiency.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laura Nadal et al.
Frontiers in aging neuroscience, 9, 24-24 (2017-02-24)
The development of the nervous system involves the overproduction of synapses but connectivity is refined by Hebbian activity-dependent axonal competition. The newborn skeletal muscle fibers are polyinnervated but, at the end of the competition process, some days later, become innervated
Patrick J O'Hern et al.
eLife, 6 (2017-05-04)
Spinal Muscular Atrophy (SMA) is caused by diminished Survival of Motor Neuron (SMN) protein, leading to neuromuscular junction (NMJ) dysfunction and spinal motor neuron (MN) loss. Here, we report that reduced SMN function impacts the action of a pertinent microRNA
Laura Nadal et al.
Molecular brain, 9(1), 67-67 (2016-06-25)
The development of the nervous system involves an initially exuberant production of neurons that make an excessive number of synaptic contacts. The initial overproduction of synapses promotes connectivity. Hebbian competition between axons with different activities (the least active are punished)
Maria Bertuzzi et al.
Scientific reports, 8(1), 1988-1988 (2018-02-02)
While cholinergic neuromodulation is important for locomotor circuit operation, the specific neuronal mechanisms that acetylcholine employs to regulate and fine-tune the speed of locomotion are largely unknown. Here, we show that cholinergic interneurons are present in the zebrafish spinal cord
Alessandra Lo Cicero et al.
Scientific reports, 6, 34798-34798 (2016-10-16)
Hutchinson-Gilford progeria syndrome (HGPS) is a rare fatal genetic disorder that causes systemic accelerated aging in children. Thanks to the pluripotency and self-renewal properties of induced pluripotent stem cells (iPSC), HGPS iPSC-based modeling opens up the possibility of access to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service