Skip to Content
Merck

Skip To

C0780

Creatine

suitable for cell culture

Synonym(s):

(α-Methylguanido)acetic acid, N-Amidinosarcosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2NC(=NH)N(CH3)CH2CO2H
CAS Number:
Molecular Weight:
131.13
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
200-306-6
MDL number:
Beilstein/REAXYS Number:
907175
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Creatine, anhydrous

grade

anhydrous

Quality Segment

form

powder

technique(s)

cell culture | mammalian: suitable

color

white

SMILES string

CN(CC(O)=O)C(N)=N

InChI

1S/C4H9N3O2/c1-7(4(5)6)2-3(8)9/h2H2,1H3,(H3,5,6)(H,8,9)

InChI key

CVSVTCORWBXHQV-UHFFFAOYSA-N

Application

Creatine (Cr) and phosphocreatine (PCr) are involved with rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). Creatine may be used as a supplement to study its uptake mechanism and metabolism of action.

Biochem/physiol Actions

Creatine is a nitrogenous compound that acts as a high-energy reservoir for the rapid regeneration of ATP.
Creatine is a nitrogenous compound that acts as a high-energy reservoir for the rapid regeneration of ATP. Approximately 95% of creatine is found in skeletal muscle, primarily as phosphocreatine. Creatine can be acquired through dietary consumption or formed from L-arginine, glycine, and L-methionine in a multi-step reaction that occurs in the kidneys and liver. Creatine is then transported to muscle tissue. Creatine supplementation is used for the enhancement of sports performance, primarily by increasing muscle mass. Creatine is also being investigated as a treatment of neuromuscular diseases, where it may aid in neuroprotection and by improving the cellular bioenergetic state.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7477A3594A0876
description

anhydrous

description

anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%

description

BioPerformance Certified, ≥99.0%, suitable for cell culture

description

-

form

powder

form

powder or crystals

form

crystalline powder

form

powder

grade

anhydrous

grade

anhydrous

grade

BioPerformance Certified

grade

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable, ligand binding assay: suitable

color

white

color

white to off-white

color

-

color

white

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Creatine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. I am going to treat the hmc3 cells with Creatine. However, I am unable to find a concentration in the papers that used this product. Could you kindly help me with this?

    1 answer
    1. A validated concentration for HMC3 cells treated with this product was not found. Creatine anhydrous is suitable for mammalian cell culture. In vitro studies using creatine report concentrations in the millimolar range. Recommend a stepwise dose–response: 0.5, 1, 2.5, 5, and 10 mM, with 5 mM as a practical starting point, accompanied by cell viability assessment.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters