Skip to Content
Merck
All Photos(1)

Documents

L134

Sigma-Aldrich

Linopirdine

≥98% (HPLC)

Synonym(s):

1,3-Dihydro-1-phenyl-3,3-bis(4-pyridinylmethyl)-2H-indol-2-one, DuP 996

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21N3O
CAS Number:
Molecular Weight:
391.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98% (HPLC)

color

white to off-white

solubility

DMSO: >10 mg/mL
ethanol: 240 mg/mL

SMILES string

O=C1N(c2ccccc2)c3ccccc3C1(Cc4ccncc4)Cc5ccncc5

InChI

1S/C26H21N3O/c30-25-26(18-20-10-14-27-15-11-20,19-21-12-16-28-17-13-21)23-8-4-5-9-24(23)29(25)22-6-2-1-3-7-22/h1-17H,18-19H2

InChI key

YEJCDKJIEMIWRQ-UHFFFAOYSA-N

Gene Information

General description

Linopiridine is a class of drugs that activates the nervous system and enhances the acetylcholine (Ach) in the brain. It can be prepared by reacting oxiindole with 4-picolyl chloride hydrochloride in the presence of a KF-Al2O3 catalyst.

Biochem/physiol Actions

Stimulates release of acetylcholine and other neurotransmitters; cognitive enhancer.

Legal Information

Sold under exclusive license from DuPont.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted rapid synthesis of neurotransmitter release enhancer linopiridine and its new analogues
Yadav JS and Reddy BVS
Synthetic Communications, 33(18), 3115-3121 (2003)
Motoharu Yoshida et al.
Journal of neurophysiology, 98(5), 2779-2794 (2007-08-31)
The M-current (current through Kv7 channels) is a low-threshold noninactivating potassium current that is suppressed by muscarinic agonists. Recent studies have shown its role in spike burst generation and intrinsic subthreshold theta resonance, both of which are important for memory
Olga Zavaritskaya et al.
Hypertension (Dallas, Tex. : 1979), 61(1), 151-159 (2012-11-28)
KCNQ channels have been identified in arterial smooth muscle. However, their role in vasoregulation and chronic vascular diseases remains elusive. We tested the hypothesis that KCNQ channels contribute to periadventitial vasoregulation in peripheral skeletal muscle arteries by perivascular adipose tissue
Richardson N Leão et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(42), 13353-13364 (2009-10-23)
While the synaptic mechanisms involved in the generation of in vitro network oscillations have been widely studied, little is known about the importance of voltage-gated currents during such activity. Here we study the role of the M-current (I(M)) in the
Alexander R Mackie et al.
The Journal of pharmacology and experimental therapeutics, 325(2), 475-483 (2008-02-15)
Pressor effects of the vasoconstrictor hormone arginine vasopressin (AVP), observed when systemic AVP concentrations are less than 100 pM, are important for the physiological maintenance of blood pressure, and they are also the basis for therapeutic use of vasopressin to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service