Skip to Content
Merck
All Photos(1)

Key Documents

SML0325

Sigma-Aldrich

Retigabine

≥98% (HPLC)

Synonym(s):

D-23129, Ezogabine, N-(2-Amino-4-(4-fluorobenzylamino)phenyl)carbamic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18FN3O2
CAS Number:
Molecular Weight:
303.33
Beilstein:
8072099
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule V

color

white to light brown

solubility

DMSO: >15 mg/mL

storage temp.

room temp

SMILES string

FC1=CC=C(CNC2=CC(N)=C(NC(OCC)=O)C=C2)C=C1

InChI

1S/C16H18FN3O2/c1-2-22-16(21)20-15-8-7-13(9-14(15)18)19-10-11-3-5-12(17)6-4-11/h3-9,19H,2,10,18H2,1H3,(H,20,21)

InChI key

PCOBBVZJEWWZFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Retigabine (Ezogabine) is a Kv7.2-7.5 (KCNQ2-5) neuronal potassium channel opener witrh anticonvulsant activity.
Retigabine is a Kv7.2-7.5 potassium channel activator; anticonvulsant.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roger J Porter et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 4(1), 149-154 (2007-01-03)
Retigabine is a novel antiseizure drug that acts through potassium channels and has activity in a broad range of animal models of epilepsy. It is also effective in several preclinical pain models. The drug has been extensively studied in phase
Martin J Gunthorpe et al.
Epilepsia, 53(3), 412-424 (2012-01-10)
The pharmacologic profile of retigabine [RTG (international nonproprietary name); ezogabine, EZG (U.S. adopted name)], is different from all currently approved antiepileptic drugs (AEDs). Its primary mechanism of action (MoA) as a positive allosteric modulator of KCNQ2-5 (K(v) 7.2-7.5) ion channels
Gökce Orhan et al.
Expert opinion on pharmacotherapy, 13(12), 1807-1816 (2012-07-13)
Epilepsy is a serious and common chronic neurological disease with an urgent need for novel treatment options, because 30% of all epilepsy patients do not respond to currently available drugs. Retigabine/Ezogabine (RTG) is a third-generation antiepileptic drug (AED) with a
Philip N Patsalos et al.
Expert opinion on pharmacotherapy, 13(5), 699-715 (2012-03-13)
The search for new, more effective antiepileptic drugs (AEDs) continues. The three most recently approved drugs, the so-called third-generation AEDs, include lacosamide, retigabine and eslicarbazepine acetate and are licensed as adjunctive treatment of partial epilepsy in adults. For the above
Michele A Faulkner et al.
Expert opinion on drug safety, 12(6), 847-855 (2013-07-26)
Complex-partial seizures are frequently resistant to antiepileptic therapy. Two new medications with mechanisms of action novel within the antiepileptic class have recently received approval for the adjunctive treatment of partial (focal) seizures. A Medline search was conducted to identify preclinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service