Skip to Content
Merck
All Photos(3)

Documents

I1006

Sigma-Aldrich

Bis(3-aminopropyl)amine

98%

Synonym(s):

BAPA, 3,3′-Diaminodipropylamine, 3,3′-Iminodipropylamine, N-(3-Aminopropyl)-1,3-propanediamine, Dipropylenetriamine, Norspermidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
131.22
Beilstein:
1071254
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.481 (lit.)

bp

151 °C/50 mmHg (lit.)

mp

−14 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

SMILES string

NCCCNCCCN

InChI

1S/C6H17N3/c7-3-1-5-9-6-2-4-8/h9H,1-8H2

InChI key

OTBHHUPVCYLGQO-UHFFFAOYSA-N

Gene Information

mouse ... Odc1(18263)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bis(3-aminopropyl)amine (BAPA) can be used:
  • To synthesize chitosan based hydrogels having potential biomedical applications.
  • For the functionalization of nanoparticles to yield terminal −NH2 groups for further modifications.
  • As an internal dispersing agent for the synthesis of water-dispersible polyurethanes.
  • As a ligand in the preparation of [Cu(bapa)]2[Mo(CN)8]·7H2O photomagnetic coordination polymer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Muta. 2 - Skin Corr. 1A - Skin Sens. 1A - STOT RE 2

Target Organs

thymus

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

242.6 °F - closed cup

Flash Point(C)

117 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photo-induced magnetic properties of the [CuII(bapa)]2[MoIV(CN) 8]?7H2O molecular ribbon.
Stefanczyk O, et al.
Journal of Material Chemistry C, 3(33), 8712-8719 (2015)
Palladium nanoparticles immobilized on EDTA-modified Fe3O4@ SiO2 nanospheres as an efficient and magnetically separable catalyst for Suzuki and Sonogashira cross-coupling reactions.
Esmaeilpour M, et al.
Applied Organometallic Chemistry, 32(4), e4302-e4302 (2018)
Isocyanate-Free Approach to Water-Borne Polyurea Dispersions and Coatings.
Ma S, et al.
ChemSusChem, 11(1), 149-158 (2018)
A simple strategy for robust preparation and characterisation of hydrogels derived from chitosan and amino functional monomers for biomedical applications
Khan F, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 6(31), 5115-5129 (2018)
T Shinki et al.
European journal of biochemistry, 183(2), 285-290 (1989-08-01)
We have reported that spermidine N1-acetyltransferase has a larger role than ornithine decarboxylase in putrescine synthesis in chick duodenum induced by 1 alpha,25-dihydroxycholecalciferol (calcitriol) [Shinki, T., Kadofuku, T., Sato, T. and Suda, T. (1986) J. Biol. Chem. 261, 11712-11716]. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service