Skip to Content
Merck
All Photos(2)

Key Documents

S3256

Sigma-Aldrich

Spermine

≥97%

Synonym(s):

N,N′-Bis(3-aminopropyl)-1,4-diaminobutane, Gerontine, Musculamine, Neuridine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH(CH2)4NH(CH2)3NH2
CAS Number:
Molecular Weight:
202.34
Beilstein:
1750791
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

biological source

microbial
synthetic

description

form: solid or semisolid

Assay

≥97%

bp

150 °C/5 mmHg (lit.)

mp

28-30 (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[H]N(CCCN)CCCCN([H])CCCN

InChI

1S/C10H26N4/c11-5-3-9-13-7-1-2-8-14-10-4-6-12/h13-14H,1-12H2

InChI key

PFNFFQXMRSDOHW-UHFFFAOYSA-N

Gene Information

human ... GRIN2B(2904)
rat ... Grin2a(24409)

Looking for similar products? Visit Product Comparison Guide

General description

Spermine is a polyamine, which functions as a free radical scavenger. It modulates gene expression, chromatin stabilization and prevents DNA damage. Spermine inhibits endonuclease-mediated DNA fragmentation.

Application

Spermine has been used:
  • as an inositol 1,4,5-trisphosphate receptor (IP3R)/glucose-regulated protein 75 (Grp75)/voltage-dependent anion channel 1 (VDAC1)/mitochondrial calcium uniporter (MCU) agonist to activate MCU in mouse podocytes
  • in the biolistic transfection of filarial parasites
  • for the production of adeno-associated virus (AAV)

Biochem/physiol Actions

Mixed NMDA agonist/antagonist at the polyamine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA glutamate receptor agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IP 3 R-Grp75-VDAC1-MCU calcium regulation axis antagonists protect podocytes from apoptosis and decrease proteinuria in an Adriamycin nephropathy rat model
Xu H, et al.
BMC Nephrology, 19(1), 140-140 (2018)
Brugia malayi: transient transfection by microinjection and particle bombardment
Higazi TB, et al.
Experimental Parasitology, 100(2), 95-102 (2002)
Using mammalian GFP reconstitution across synaptic partners (mGRASP) to map synaptic connectivity in the mouse brain.
Feng L, et al.
Nature Protocols, 9(10), 2425-2425 (2014)
H C Ha et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(19), 11140-11145 (1998-09-16)
The polyamines are small organic cations that are absolutely required for eukaryotic cell growth. Although their growth requirements are well established, the molecular functions of the polyamines are ill-defined. Oxidative damage to DNA by reactive oxygen species is a continual
H Hosseinkhani et al.
Gene therapy, 11(2), 194-203 (2004-01-09)
Dextran-spermine cationic polysaccharide was prepared by means of reductive amination between oxidized dextran and the natural oligoamine spermine. The formed Schiff-base imine-based conjugate was reduced with borohydride to obtain the stable amine-based conjugate. The transfection efficiency of the synthetic dextran-spermine

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service