
Bis(3-aminopropyl)amine

Synonym(s):
3,3′-Diaminodipropylamine, 3,3′-Iminodipropylamine, N-(3-Aminopropyl)-1,3-propanediamine, Dipropylenetriamine, Norspermidine
Linear Formula:
(NH2CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
131.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing