Skip to Content
Merck
All Photos(2)

Documents

GF64339724

Cobalt

foil, not light tested, 100x100mm, thickness 0.008mm, 99.9%

Synonym(s):

Cobalt, CO000130

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

form

foil

manufacturer/tradename

Goodfellow 643-397-24

resistivity

6.24 μΩ-cm, 20°C

size × thickness

100x100 mm × 0.008 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Nayak et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 54(2), 189-193 (2001-02-24)
Heavy ion activation of natural cobalt foil with 52 MeV 11B4+ and 73 MeV 12C6+ results in the formation of carrier-free (66,67)Ge and (66,67)As radionuclides, respectively, along with their corresponding decay products, (66,67)Ga in the matrix. The carrier free gallium
Dalia Nayak et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 65(8), 891-896 (2007-05-29)
Heavy ion activation of natural cobalt foil with 84MeV (12)C results in the formation of no-carrier-added (nca) (66,67)As radionuclides, along with their corresponding decay products, (66,67)Ge and (66,67)Ga, in the matrix. Because arsenic and germanium radionuclides are short-lived, after a
William L Griffin
The Journal of arthroplasty, 29(4), 659-660 (2014-03-25)
Ion levels have been shown to reliably predict abnormal function of the bearing surface with increased wear, but ion levels should not be used alone as a trigger for when to proceed with revision surgery with metal-metal articulations. Risk stratification
Anthony F Pizon et al.
Journal of medical toxicology : official journal of the American College of Medical Toxicology, 9(4), 416-417 (2013-11-22)
Prosthetic hip-associated cobalt toxicity (PHACT) is gaining recognition due to the use of metal-on-metal total hip replacements. Identifying true toxicity from merely elevated cobalt levels can be extremely difficult due to the lack of available data. An extensive review of
Hao-Di Wu et al.
Journal of nanoscience and nanotechnology, 14(6), 4097-4100 (2014-04-18)
Cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane were prepared by a facile solution method and fully characterized by SEM, AFM, XRD, Raman, EDX, and UV-vis-NIR. The as-prepared cocrystal nanofibres had smooth surfaces and uniform dimension. When incorporated into prototype devices

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service