Merck
Prosthetic hip-associated cobalt toxicity.
Journal of medical toxicology : official journal of the American College of Medical Toxicology (2013-11-22)
Anthony F Pizon, Michael Abesamis, Andrew M King, Nathan Menke
ABSTRACT
Prosthetic hip-associated cobalt toxicity (PHACT) is gaining recognition due to the use of metal-on-metal total hip replacements. Identifying true toxicity from merely elevated cobalt levels can be extremely difficult due to the lack of available data. An extensive review of the medical literature was undertaken to characterize cobalt toxicity from prosthetic hips. As an objective approach to making the diagnosis of PHACT, we suggest the following criteria: (1) elevated serum or whole blood cobalt levels due to a prosthetic hip, (2) at least two test-confirmed findings consistent with cobalt toxicity, and (3) exclusion of other etiologies. Adhering to objective diagnostic data for PHACT is a realistic and prudent method by which to eliminate the subjectivity of vague or difficult to identify complaints. These diagnostic criteria are not meant to evaluate prosthetic hardware failure, but as a means to identify systemic cobalt toxicity. Finally, assessment of cobalt toxicity from prosthetic hips should be done in conjunction with a medical toxicologist.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Cobalt, wire reel, 1m, diameter 0.1mm, hard, 99.99+%
Cobalt, wire reel, 2m, diameter 0.1mm, hard, 99.99+%
Cobalt, wire reel, 1m, diameter 0.25mm, hard, 99.99+%
Cobalt, wire reel, 1m, diameter 0.5mm, hard, 99.99+%
Cobalt, wire reel, 0.2m, diameter 2.0mm, hard, 99.99+%
Cobalt, wire reel, 0.5m, diameter 0.125mm, hard, 99.99+%
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Cobalt, wire reel, 1m, diameter 0.05mm, hard, 99.9%
Cobalt, wire reel, 0.2m, diameter 0.05mm, hard, 99.9%
Cobalt, wire reel, 0.5m, diameter 0.1mm, hard, 99.99+%
Cobalt, wire reel, 2m, diameter 0.125mm, hard, 99.99+%
Cobalt, wire reel, 0.1m, diameter 0.05mm, hard, 99.9%
Cobalt, wire reel, 0.5m, diameter 0.05mm, hard, 99.9%
Cobalt, wire reel, 0.2m, diameter 0.125mm, hard, 99.99+%
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, 15mm disks, thickness 0.02mm, 99.9%
Cobalt, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Cobalt, wire reel, 0.2m, diameter 0.1mm, hard, 99.99+%
Cobalt, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cobalt, wire reel, 0.5m, diameter 1.0mm, hard, 99.99+%