- Preparation of cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane with good photoresponse.
Preparation of cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane with good photoresponse.
Journal of nanoscience and nanotechnology (2014-04-18)
Hao-Di Wu, Yan Xiao, Feng-Xia Wang, Ge-Bo Pan
PMID24738357
ABSTRACT
Cocrystal nanofibres of cobalt octaethylporphyrin and tetracyanoquinodimethane were prepared by a facile solution method and fully characterized by SEM, AFM, XRD, Raman, EDX, and UV-vis-NIR. The as-prepared cocrystal nanofibres had smooth surfaces and uniform dimension. When incorporated into prototype devices, they exhibited good photoresponse at ambient conditions. Additionally, the phototransistor characteristics with a maximum I(on)/I(off) ratio of -460 was demonstrated. The facile synthesis and good photoresponse may boost the potential applications of cocrystal-based nanostructures in future miniaturized devices.
Product Description
Cobalt, foil, 8mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.002mm, temporary acrylic support, 99.9%
Cobalt, foil, 8mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 8mm disks, thickness 0.005mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.0125mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Cobalt, microfoil, disks, 10mm, thinness 0.5μm, specific density 434μg/cm2, permanent mylar 3.5μm support, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Cobalt, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.9%
Cobalt, wire reel, 0.1m, diameter 1.0mm, hard, 99.99+%
Cobalt, foil, 8mm disks, thickness 0.003mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.0125mm, 99.9%