Skip to Content
Merck
All Photos(2)

Key Documents

D100951

Sigma-Aldrich

N,N-Diethyl-3-methylbenzamide

97%

Synonym(s):

N,N-Diethyl-m-toluamide, m-Toluic acid diethylamide, DEET

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CON(C2H5)2
CAS Number:
Molecular Weight:
191.27
Beilstein:
2046711
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.7 (vs air)

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.523 (lit.)

bp

111 °C/1 mmHg
160 °C/19 mmHg (lit.)

density

0.998 g/mL at 20 °C (lit.)

SMILES string

CCN(CC)C(=O)c1cccc(C)c1

InChI

1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3

InChI key

MMOXZBCLCQITDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Diethyl-3-methylbenzamide can undergo reduction in the presence of iron catalyst to synthesize N,N-diethyl-3-methylbenzamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasan Soliman Yusufoglu et al.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society, 26(6), 860-864 (2018-09-12)
The essential oils (EOs) of Anthemis melampodina (Am) and Anthemis scrobicularis (As) (Asteraceae) were extracted from the aerial parts of the plants by hydrodistillation, and their chemical compositions were analyzed using GC-FID and GC-MS. Fifty-six components representing 85.5% of the
The insect repellent DEET (N, N-diethyl-3-methylbenzamide) increases the synthesis of glutathione S-transferase in cultured mosquito cells.
Hellestad VJ, et al.
Cell Biology and Toxicology, 27(2), 149-157 (2011)
A Convenient and General Iron-Catalyzed Reduction of Amides to Amines.
Zhou S, et al.
Angewandte Chemie (International Edition in English), 121(50), 9671-9674 (2009)
Melanie Ramírez et al.
Parasites & vectors, 13(1), 142-142 (2020-03-20)
Studying the behavioral response of blood-sucking disease-vector insects to potentially repellent volatile compounds could shed light on the development of new control strategies. Volatiles released by human facial skin microbiota play different roles in the host-seeking behavior of triatomines. We
Ivanka B Semerdjieva et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-20)
Satureja pilosa Velen. senso lato is a Balkan endemic plant that is not well characterized and is found on rocky outcrops of limestone base in Stara Planina (the Balkan Mountains) and the Rhodope Mountains. The objective of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service