Skip to Content
Merck
All Photos(1)

Key Documents

202320

Sigma-Aldrich

Poly(propylene glycol)

average Mn ~1,000

Synonym(s):

PPG, Poly(propylene oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OCH(CH3)CH2]nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

mol wt

average Mn ~1,000

contains

130-190 ppm proprietary phenolic antioxidant

refractive index

n20/D 1.449

viscosity

150 cSt(25 °C)(lit.)

hydroxyl value

111 mg KOH/g

solubility

water: miscible (completely)

density

1.005 g/mL at 25 °C

SMILES string

CC(O)CO

InChI

1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3

InChI key

DUFKCOQISQKSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(propylene glycol) (PPG) is a biocompatible polymer widely used as a precursor to fabricate biomedical materials. It is soluble in water at low temperatures and becomes insoluble at high temperatures.

Application

Poly(propylene glycol) can be used:
  • As a precursor to synthesize multiblock thermo-responsive smart polymers for various biomedical applications. The presence of PPG is the main reason for the thermosensitivity of these polymers. For example, it can be used to synthesize poly(ester urethane)s.
  • For the surface modification of sisal fiber to enhance nucleating ability.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

445.0 °F - closed cup

Flash Point(C)

229.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Chin Wu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(8), 2232-2243 (2016-07-23)
Despite its common usage in vertebral augmentation procedures (VAPs), shortcomings of commercial polymethylmethacrylate (PMMA) still remain. Accordingly, injectable and biodegradable composite cements, which are composed of poly(propylene fumarate)/α-tricalcium/hydroxyapatite (PPF/α-TCP/HAP) and PPF/tetracalcium phosphate/dicalcium phosphate (PPF/TtCP/DCP), were developed. A porcine model was
Izzuddin Zaman et al.
Nanoscale, 4(15), 4578-4586 (2012-06-19)
In spite of extensive studies conducted on carbon nanotubes and silicate layers for their polymer-based nanocomposites, the rise of graphene now provides a more promising candidate due to its exceptionally high mechanical performance and electrical and thermal conductivities. The present
R De Lisi et al.
Physical chemistry chemical physics : PCCP, 13(27), 12571-12577 (2011-06-15)
The study highlighted the main forces driving the formation of hydroxypropyl-cyclodextrins (HP-CDs) + poly(propylene) glycol 725 g mol(-1) inclusion complexes. The temperature parameter was chosen as the variable to modulate the hydrophobicity of the polymer, and consequently ITC experiments as
Mohammed Taghi Zafarani-Moattar et al.
Biotechnology progress, 28(1), 146-156 (2011-09-29)
In this work, we proposed a novel aqueous biphasic system (ABS) composed of polypropylene glycol P400 (PPG P400) and hydrophilic ionic liquids (IL), 1-alkyl-3-methylimidazolium bromide (alkyl = ethyl or butyl), forming an upper polymer-rich phase and a lower IL-rich phase
L Hong et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 83(6 Pt 1), 061508-061508 (2011-07-30)
There are various arguments and models connecting the characteristic length associated with the boson peak vibrations ξ to the length scale of dynamical heterogeneity L(het). ξ is usually defined as the ratio of the transverse sound velocity to the boson

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service