Skip to Content
Merck
All Photos(1)

Key Documents

A34805

Sigma-Aldrich

2-Allylphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H4OH
CAS Number:
Molecular Weight:
134.18
Beilstein:
742121
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.546 (lit.)

bp

220 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1CC=C

InChI

1S/C9H10O/c1-2-5-8-6-3-4-7-9(8)10/h2-4,6-7,10H,1,5H2

InChI key

QIRNGVVZBINFMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Ye Hu et al.
Journal of environmental sciences (China), 17(3), 491-493 (2005-08-09)
The method of residue analysis of a new synthesized fungicide 2-allylphenol was studied by simulating the active compound structure in Gingko tree (Gingko biloba L.) and its dissipation rate and terminal residue levels in tomato under field condition. Residues of
Alvin Kah-wei Hee et al.
Journal of chemical ecology, 30(11), 2127-2138 (2005-01-28)
Pharmacophagy of methyl eugenol (ME)--a highly potent male attractant, by Bactrocera papayae results in the hydroxylation of ME to sex pheromonal components, 2-ally-4,5-dimethoxyphenol (DMP) and (E)-coniferyl alcohol (CF). These compounds, which are also male attractants, are then sequestered and stored
Shuangjun Gong et al.
Pest management science, 65(12), 1337-1343 (2009-08-18)
2-Allylphenol is a registered fungicide in China to control fungal diseases on tomato, strawberry and apple. It is synthetic and structurally resembles the active ingredient ginkgol isolated from Ginkgo biloba L. bark. 2-Allylphenol has been used in China for 10
Meyyappan Arumugam et al.
Applied microbiology and biotechnology, 86(1), 109-117 (2009-11-11)
A marine actinobacterium isolated from the Bay of Bengal, India and previously found to be producing an antimicrobial and cytotoxic terpenoid was further investigated for antimicrobial metabolites. The bacterium was preliminarily identified as a new species of the genus Streptomyces
Tianli Qu et al.
Ecotoxicology and environmental safety, 102, 136-141 (2014-02-18)
2-Allylphenol is a biomimetic synthetic fungicide that mimics the compound ginkgol found in gingko fruit (Gingko biloba L.). This systemic fungicide can effectively suppress a wide range of plant diseases, including wheat sharp eyespot (Rhizoctonia cerealis). However, its degradation in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service