Skip to Content
Merck
All Photos(2)

Key Documents

E51791

Sigma-Aldrich

Eugenol

ReagentPlus®, 99%

Synonym(s):

2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2C=CHCH2)C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1366759
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.541 (lit.)

bp

254 °C (lit.)

mp

−12-−10 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

COc1cc(CC=C)ccc1O

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3

InChI key

RRAFCDWBNXTKKO-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

Eugenol is a natural phenolic compound found in clove oil.[1]

Application

Eugenol can be used in the synthesis of:
  • bio-based benzoxazines[1]
  • ibuprofen eugenol ester (IEE)[2]
  • polycarbonate/cyanate ester blends[3]
  • 1,2,3-triazole eugenol glucosides[4]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antifungal efficacy of thymol, carvacrol, eugenol and menthol as alternative agents to control the growth of food-relevant fungi.
Abbaszadeh S, et al.
Journal de mycologie medicale, 24(2), e51-e56 (2014)
Synthesis and characterization of novel bio-based benzoxazines from eugenol
Thirukumaran P, et al.
Royal Society of Chemistry Advances, 4(16), 7959-7966 (2014)
Poly (DL?lactide?co?glycolide)(PLGA) nanoparticles with entrapped trans?cinnamaldehyde and eugenol for antimicrobial delivery applications.
Gomes C, et al.
Journal of Food Science, 76(2) (2011)
Synthesis and antimicrobial activity of 6-triazolo-6-deoxy eugenol glucosides
de Souza TB, et al.
Carbohydrate Research, 410(20), 1-8 (2015)
Synthesis and characterization of a renewable cyanate ester/polycarbonate network derived from eugenol
Harvey BG, et al.
Polymer, 55(20), 5073-5079 (2014)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the concentration of Eugenol? I've purchased Eugenol E51791 5G, but it's concentration is not written. How many grames in how many ml?

    1 answer
    1. Eugenol is a neat liquid with a melting point of −12-−10 °C. The density of this material is 1.067 g/mL. The 5G pack size will equate to 4.69 mL, the 100G pack size will equate to 93.72 mL, and the 500 G pack size will equate to 468.60 mL. Based on the minimum 99% purity this product is approx. 6.4 M.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service