Skip to Content
Merck
All Photos(3)

Key Documents

441090

Sigma-Aldrich

2,2′-Azobis(2-methylpropionitrile)

98%

Synonym(s):

α,α′-Azoisobutyronitrile, AIBN, Azobisisobutyronitrile, Free radical initiator

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(CN)N=NC(CH3)2CN
CAS Number:
Molecular Weight:
164.21
Beilstein:
1708400
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

102-104 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(\N=N\C(C)(C)C#N)C#N

InChI

1S/C8H12N4/c1-7(2,5-9)11-12-8(3,4)6-10/h1-4H3/b12-11+

InChI key

OZAIFHULBGXAKX-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-Azobis(2-methylpropionitrile) can be used as an initiator in the preparation of:
  • Polystyrene by soap-free emulsion polymerization.
  • Molecularly imprinted polymer(MIP) using 1-vinyl imidazole. MIP can be used to quantify acid violet 19 dye in river water samples.

Storage and Stability

Warning: these products are subject to the Explosives Act and must be transported refrigerated - additional costs for transport will apply.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Self-react. C

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Luna Quinto et al.
Journal of hazardous materials, 384, 121374-121374 (2019-11-02)
A molecularly imprinted polymer (MIP) was developed for the determination of acid violet 19 (AV19) dye. The MIP was synthesized by polymerization using 1-vinyl imidazole (functional monomer) and 2,2'-azobis(2-methylpropionitrile) as the radical initiator. The functional monomer was previously selected by
Tetrahedron Letters, 48, 5585-5585 (2007)
Influence of the size of polystyrene synthesized through soap-free emulsion polymerization on antimicrobial activity
Tetsuya Yamamoto, et al.
Materials Today Communications, 20, 100572-100572 (2019)
Mathieu Le Noir et al.
Journal of separation science, 32(9), 1471-1479 (2009-04-29)
Highly efficient removal of endocrine-disrupting compounds (EDCs) such as 17beta-estradiol (E2), 4-nonylphenol (NP) and atrazine from water was achieved using a novel macroporous adsorption medium. The medium consisted of a macroporous poly(vinyl alcohol) (PVA) cryogel with molecularly imprinted polymer (MIP)
K Shivaji Sharma et al.
Langmuir : the ACS journal of surfaces and colloids, 24(23), 13581-13590 (2008-11-05)
A novel class of nonionic amphipols (NAPols) designed to handle membrane proteins in aqueous solutions has been synthesized, and its solution properties have been examined. These were synthesized through free radical cotelomerization of glucose-based hydrophilic and amphiphilic monomers derived from

Articles

To keep pace with Moore′s Law, there is a continuing need in the semiconductor industry to achieve higher circuit density in microelectronic devices.

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

An article regarding common FAQs for initiators and stabilizers.

See All

Protocols

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

RAFT polymerization offers precise control, enabling tailored synthesis of complex polymer structures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service