Skip to Content
Merck
All Photos(3)

Key Documents

852678

Sigma-Aldrich

6-Mercaptopurine monohydrate

98%

Synonym(s):

6-Purinethiol, 6-Thiohypoxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4S · H2O
CAS Number:
Molecular Weight:
170.19
Beilstein:
4012091
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

O.Sc1ncnc2[nH]cnc12

InChI

1S/C5H4N4S.H2O/c10-5-3-4(7-1-6-3)8-2-9-5;/h1-2H,(H2,6,7,8,9,10);1H2

InChI key

WFFQYWAAEWLHJC-UHFFFAOYSA-N

Gene Information

human ... PPAT(5471)

Looking for similar products? Visit Product Comparison Guide

General description

6-Mercaptopurine monohydrate is an antimetabolite and antineoplastic drug. Its cocrystallization with 4-hydroxybenzoic acid and 2,4-dihydroxybenzoic acid or salt formation with piperazine may improve its aqueous solubility.

Application

6-Mercaptopurine is a widely used antileukemic agent that inhibits de novo purine synthesis through incorporation of thiopurine methyltransferase metabolites into DNA and RNA.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving the solubility of 6-Mercaptopurine via cocrystals and salts
Xu LL, et al.
Crystal Growth & Design, 12(12), 6004-6011 (2012)
The crystal and molecular structure of 6-mercaptopurine monohydrate.
Sletten E, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 25(7), 1330-1338 (1969)
Comparative structural and spectrum analyses of 6-mercaptopurine monohydrate and 6-mercaptopurine hydrochloride
Perez-Ruiz E, et al.
Canadian Journal of Analytical Sciences and Spectroscopy, 43(3), 59-67 (1998)
Unit-cell dimensions and space group of 6-mercaptopurine monohydrate.
Hoogsteen K.
Nature, 178(4529), 379-379 (1956)
Goran Marinković et al.
Journal of immunology (Baltimore, Md. : 1950), 192(9), 4370-4378 (2014-03-29)
Azathioprine and its metabolite 6-mercaptopurine (6-MP) are well established immunosuppressive drugs. Common understanding of their immunosuppressive properties is largely limited to immune cells. However, in this study, the mechanism underlying the protective role of 6-MP in endothelial cell activation is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service