Skip to Content
Merck
All Photos(1)

Key Documents

769304

Sigma-Aldrich

Barium peroxide

anhydrous, ≥86.0% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaO2
CAS Number:
Molecular Weight:
169.33
EC Number:
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

≥86.0% (RT)

form

powder

loss

≤0.1% loss on drying

anion traces

chloride (Cl-): ≤0.05%

cation traces

Fe: ≤0.02%
heavy metals (as Pb): ≤0.01%

SMILES string

[Ba++].[O-][O-]

InChI

1S/Ba.O2/c;1-2/q+2;-2

InChI key

ZJRXSAYFZMGQFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Barium peroxide is a precursor material used for synthesis of novel magnetic and superconducting oxides. It also finds application in thermochemical energy storage systems.

Analysis Note

Substances insoluble in dilute hydrochlorid acid ≤ 1.5 %
Substances not precipitated by diluted sulfuric acid (as sulphates) ≤ 2.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ba2IrO4: A spin-orbit Mott insulating quasi-two-dimensional antiferromagnet
Okabe, H., et al.
Physical Review. B, Condensed Matter and Materials Physics, 83, 155118-155118 (2011)
A J Carrillo et al.
Physical chemistry chemical physics : PCCP, 18(11), 8039-8048 (2016-03-01)
The barium peroxide-based redox cycle was proposed in the late 1970s as a thermochemical energy storage system. Since then, very little attention has been paid to such redox couples. In this paper, we have revisited the use of reduction-oxidation reactions
Honda, R., et al.
Physica C, 426, 613-617 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service