Skip to Content
Merck
All Photos(3)

Key Documents

466271

Sigma-Aldrich

Calcium peroxide

75%, −200 mesh

Synonym(s):

Calcium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaO2
CAS Number:
Molecular Weight:
72.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

75%

form

powder

reaction suitability

reagent type: oxidant

particle size

−200 mesh

mp

>365 °C

SMILES string

[Ca++].[O-][O-]

InChI

1S/Ca.O2/c;1-2/q+2;-2

InChI key

LHJQIRIGXXHNLA-UHFFFAOYSA-N

General description

Calcium peroxide (CP) is a peroxide compound that releases oxygen and hydrogen peroxide. It is a safe inorganic peroxy compound in a solid form of oxygen. It can be used in the remediation of contaminated sites. It has the ability to construct a permeable reactive barrier system by acting as oxygen releasing material (OSM).

Application

CP can be used as an amendment that facilitates the supplying of oxygen for agriculture, horticulture and biotechnology applications.

Other Notes

Remainder calcium hydroxide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seonhee Kim et al.
The Science of the total environment, 762, 143124-143124 (2020-11-01)
Arsenic (As) is uptaken more readily by rice over wheat and barley. The exposure of As to humans being in the rice-consuming regions is a serious issue. Thus, an effective practice to reduce the translocation of As from soil to
Rijuta Ganesh Saratale et al.
Bioresource technology, 258, 26-32 (2018-03-09)
This study focuses on development of calcium peroxide (CaO2) pretreatment that removes major part of lignin but retaining most of sugar components of kenaf core powder (KCP) biomass. In chemical pretreatment, usually higher loss of biomass occurs which was less
Venu Shanmugavel et al.
Food chemistry, 311, 125964-125964 (2019-12-23)
Potassium bromate, is an oxidizing agent and one of the best and cheapest dough improvers in the baking industry. Due to its positive effects it plays a major role in the bread-making industry. Potassium bromate has significant effect on food
Oxygen release kinetics from solid phase oxygen in Arctic Alaska
Schmidtke T, et al.
Journal of Hazardous Materials, 64(2), 157-165 (1999)
Tian-Jiao Zhou et al.
Journal of controlled release : official journal of the Controlled Release Society, 309, 82-93 (2019-07-26)
Hypoxia is a potent tumor microenvironmental (TME) factor promoting immunosuppression and metastatic progression. For current anticancer therapeutic strategies, the combination of hypoxia alleviation and photodynamic therapy (PDT) might be a useful approach to further improve anticancer efficacy. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service