Skip to Content
Merck
All Photos(1)

Key Documents

689963

Sigma-Aldrich

Ethyl benzoylacetate

Arxada quality, ≥98% (GC)

Synonym(s):

Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COOC2H5
CAS Number:
Molecular Weight:
192.21
Beilstein:
389944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.6 (vs air)

Assay

≥98% (GC)

form

liquid

quality

Arxada quality

manufacturer/tradename

Arxada AG

refractive index

n20/D 1.52 (lit.)

bp

265-270 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1

InChI

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl benzoylacetate is a general reagent used in the synthesis of substituted pyrroles, oxazoles, indoles, imidazo[1, 2-a] pyridines and a variety of other aromatic compounds.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Facile synthesis of polysubstituted oxazoles via a copper-catalyzed tandem oxidative cyclization.
Wan C, et al.
Organic Letters, 12(10), 2338-2341 (2010)
Facile access to polysubstituted indoles via a cascade Cu-catalyzed arylation? condensation process.
Chen Y, et al.
The Journal of Organic Chemistry, 72(24), 9329-9334 (2007)
Expedient synthesis of highly substituted pyrroles via tandem rearrangement of α-diazo oxime ethers.
Jiang Y, et al.
Journal of the American Chemical Society, 134(9), 4104-4107 (2012)
Rhenium-and manganese-catalyzed synthesis of aromatic compounds from 1, 3-dicarbonyl compounds and alkynes.
Kuninobu Y, et al.
The Journal of Organic Chemistry, 75(2), 334-341 (2009)
TBAI-catalyzed oxidative coupling of aminopyridines with β-keto esters and 1, 3-diones?synthesis of imidazo [1, 2-a] pyridines.
Ma L, et al.
Chemical Communications (Cambridge, England), 47(40), 11333-11335 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service