Skip to Content
Merck
All Photos(1)

Key Documents

12980

Sigma-Aldrich

Ethyl benzoylacetate

redist., ≥97.0% (HPLC)

Synonym(s):

Benzoylacetic acid ethyl ester, Ethyl 3-oxo-3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH2COOC2H5
CAS Number:
Molecular Weight:
192.21
Beilstein:
389944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.6 (vs air)

Quality Level

Assay

≥97.0% (HPLC)

quality

redist.

refractive index

n20/D 1.52 (lit.)
n20/D 1.531

bp

265-270 °C (lit.)

solubility

H2O: insoluble
alcohol: miscible
diethyl ether: miscible

density

1.11 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)c1ccccc1

InChI

1S/C11H12O3/c1-2-14-11(13)8-10(12)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

GKKZMYDNDDMXSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl benzoylacetate (Benzoylacetic acid ethyl ester) was used to prepare ethyl 2-fluoro-2-benzolyacetate. It was also used to synthesize iodonium ylides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsugio Kitamura et al.
Organic letters, 13(9), 2392-2394 (2011-04-09)
A simple, practical, and convenient fluorination of 1,3-dicarbonyl compounds was achieved by direct use of aqueous hydrofluoric acid and iodosylbenzene (PhIO). The reaction of ethyl benzoylacetate with the reagent system of aqueous HF and PhIO in CH(2)Cl(2) gave ethyl 2-fluoro-2-benzolyacetate
Keisuke Gondo et al.
Molecules (Basel, Switzerland), 17(6), 6625-6632 (2012-06-26)
Reaction of dibenzoylmethane with (diacetoxyiodo)benzene in the presence of KOH in MeCN quantitatively gave the corresponding iodonium ylide, which was treated with a HF reagent to afford the corresponding 2-fluorinated dibenzoylmethane in 14-50% yields. The similar reaction of the iodonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service