Skip to Content
Merck

Skip To

688983

Ethyl acetoacetate

Arxada quality, ≥99.0% (GC)

Synonym(s):

Acetoacetic ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
205-516-1
Beilstein/REAXYS Number:
385838
MDL number:
Assay:
≥99.0% (GC)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

4.48 (vs air)

Quality Segment

vapor pressure

1 mmHg ( 28.5 °C)

assay

≥99.0% (GC)

form

liquid

autoignition temp.

580 °F

quality

Arxada quality

expl. lim.

9.5 %

manufacturer/tradename

Arxada AG

impurities

≤0.05% water, ≤0.10% acid (as acetic acid)

color

APHA: ≤15

bp

180 °C, 181 °C (lit.)

mp

−43 °C (lit.)

solubility

water: soluble 130 g/L at 20 °C

density

1.029 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Application

Ethyl acetoacetate can undergo:
  • Transesterification with primary and secondary alcohols in the presence of boric acid.
  • Microbial reduction to form (S)-ethyl 3-hydroxybutyrate.
  • C-acylation by acid chlorides in the presence of magnesium chloride and pyridine.
  • Asymmetric biphasic catalytic hydrogenation in the presence of a water-soluble ruthenium-BINAP complex.


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W24151263954537349
assay

≥99.0% (GC)

assay

≥97%

assay

≥99.0% (GC)

assay

99%

solubility

water: soluble 130 g/L at 20 °C

solubility

water: soluble 35 part, organic solvents: soluble

solubility

water: soluble 130 g/L at 20 °C

solubility

water: soluble 130 g/L at 20 °C

Quality Level

200

Quality Level

300, 400

Quality Level

100

Quality Level

200

mp

−43 °C (lit.)

mp

−43 °C (lit.)

mp

−43 °C (lit.)

mp

−43 °C (lit.)

form

liquid

form

-

form

-

form

-

bp

180 °C, 181 °C (lit.)

bp

181 °C (lit.)

bp

181 °C (lit.)

bp

181 °C (lit.)


Still not finding the right product?

Explore all of our products under Ethyl acetoacetate


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

164.3 °F - closed cup

flash_point_c

73.5 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters