Skip to Content
Merck
All Photos(2)

Key Documents

678163

Sigma-Aldrich

1,2-Dimethoxy-1,1,2,2-tetramethyldisilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3O)Si(CH3)2)2
CAS Number:
Molecular Weight:
178.38
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.425

density

0.8587 g/mL at 25 °C

SMILES string

CO[Si](C)(C)[Si](C)(C)OC

InChI

1S/C6H18O2Si2/c1-7-9(3,4)10(5,6)8-2/h1-6H3

InChI key

CWGBHCIGKSXFED-UHFFFAOYSA-N

Application

1,2-Dimethoxy-1,1,2,2-tetramethyldisilane can be used:
  • In the palladium catalyzed bis(silylation) of 1,4-diethynylbenzene to obtain a crack free sol-gel.
  • To prepare dimethylsilylene via gas-phase flow-pyrolysis at 600 0C.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.8 °F - closed cup

Flash Point(C)

27.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transition metal catalyzed element-element′ additions to alkynes
Ansell MB, et al.
Coordination Chemistry Reviews, 336, 54-77 (2017)
Tetramethylsilacyclopropene
Conlin RT and Gaspar PP
Journal of the American Chemical Society, 98(12), 3715-3716 (1976)

Articles

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Pd-catalyzed cross-coupling of silicon compounds offers an alternative in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service