Skip to Content
Merck
All Photos(2)

Key Documents

390143

Sigma-Aldrich

Triethoxysilane

95%

Synonym(s):

Silicon triethoxide, Triethoxysilyl hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3SiH
CAS Number:
Molecular Weight:
164.27
Beilstein:
1738989
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.377 (lit.)

bp

134-135 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

[H][Si](OCC)(OCC)OCC

InChI

1S/C6H16O3Si/c1-4-7-10(8-5-2)9-6-3/h10H,4-6H2,1-3H3

InChI key

QQQSFSZALRVCSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethoxysilane is an organosilicon compound used as a reducing agent for carbonyl groups and in the hydrosilylation of carbon-carbon multiple bonds.

Application

Triethoxysilane can be used as a reducing agent in the ionic liquid-catalyzed reductive amination of xylose to furfurylamines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Àngela Ribes et al.
Scientific reports, 6, 38649-38649 (2016-12-08)
We present herein the use of nanoporous anodic alumina (NAA) as a suitable support to implement "molecular gates" for sensing applications. In our design, a NAA support is loaded with a fluorescent reporter (rhodamine B) and functionalized with a short
Synthesis, 558-558 (1981)
Jianxiong Chan et al.
Sensors (Basel, Switzerland), 19(16) (2019-08-17)
Pre-treatment screening of individuals for human leukocyte antigens (HLA) HLA-B*57:01 is recommended for the prevention of life-threatening hypersensitivity reactions to abacavir, a drug widely prescribed for HIV treatment. However, the implementation of screening in clinical practice is hindered by the
Teng-Fei Fan et al.
Nature communications, 11(1), 1449-1449 (2020-03-21)
Pollen's practically-indestructible shell structure has long inspired the biomimetic design of organic materials. However, there is limited understanding of how the mechanical, chemical, and adhesion properties of pollen are biologically controlled and whether strategies can be devised to manipulate pollen
Ana R Bastos et al.
Sensors (Basel, Switzerland), 18(3) (2018-03-15)
The development of portable low-cost integrated optics-based biosensors for photonics-on-a-chip devices for real-time diagnosis are of great interest, offering significant advantages over current analytical methods. We report the fabrication and characterization of an optical sensor based on a Mach-Zehnder interferometer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service