Skip to Content
Merck
All Photos(1)

Key Documents

235733

Sigma-Aldrich

1,1,3,3-Tetramethyldisiloxane

97%

Synonym(s):

1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, Bis(dimethylsilyl)ether, Bis(dimethylsilyl)oxide, Tetramethyldisiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2SiH]2O
CAS Number:
Molecular Weight:
134.32
Beilstein:
1733364
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.370 (lit.)

bp

70-71 °C (lit.)

density

0.76 g/mL at 25 °C (lit.)

SMILES string

C[SiH](C)O[SiH](C)C

InChI

1S/C4H14OSi2/c1-6(2)5-7(3)4/h6-7H,1-4H3

InChI key

NVYQDQZEMGUESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,3,3-Tetramethyldisiloxane is an inexpensive, nontoxic, and mild reducing agent. It is an attractive hydride source for hydrosilylation reactions, dehydrogenative silylations, and polymer production due to its electrophilic nature.

Application

1,1,3,3-Tetramethyldisiloxane can be used as:
  • A reactant to synthesize mono-functionalized siloxane derivatives via hydrosilylation reaction.
  • A reducing agent to synthesize alkyl halides from aldehydes.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-9.99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient catalytic and solvent-free method for the synthesis of mono-organofunctionalized 1, 1, 3, 3-tetramethyldisiloxane derivatives
Januszewski R, et al.
Journal of Organometallic Chemistry, 846, 263-268 (2017)
1, 1, 3, 3-Tetramethyldisiloxane
Deng J
Synlett, 22, 2102-2103 (2011)
Tetrahedron Letters, 25, 3123-3123 (1984)
1, 1, 3, 3-Tetramethyldisiloxane
Laval S, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service