Skip to Content
Merck
All Photos(1)

Key Documents

469300

Sigma-Aldrich

Hexamethyldisiloxane

viscosity 0.65 cSt (25 °C)

Synonym(s):

Poly(dimethylsiloxane), HMDSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOSi(CH3)3
CAS Number:
Molecular Weight:
162.38
Beilstein:
1736258
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

refractive index

n20/D 1.377 (lit.)

viscosity

0.65 cSt(25 °C)

bp

101 °C (lit.)

mp

−59 °C (lit.)

density

0.764 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)C

InChI

1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3

InChI key

UQEAIHBTYFGYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Superhydrophobic Surface Coating: Studies the application of hexamethyldisiloxane coatings on fabric masks via plasma treatment to achieve superhydrophobic properties, enhancing the masks′ resistance to moisture (M Prado et al., 2022).
  • PECVD for Thin Film Coatings: Describes the use of hexamethyldisiloxane in plasma-enhanced chemical vapor deposition to create thin film coatings, exploring the effects of using extremely asymmetric capacitive RF discharge (Ž Gosar et al., 2020).
  • QCM-Based VOC Sensors: Hexamethyldisiloxane thin films are used as sensitive coatings for quartz crystal microbalance sensors, enhancing their capability to detect various volatile organic compounds (M Boutamine et al., 2014).
  • Hydrophobic-Hydrophilic Films: Explores the polymerization of hexamethyldisiloxane using atmospheric pressure plasma jets to create films with tailored hydrophobic and hydrophilic properties (M Bashir, S Bashir, 2015).

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan N Thon et al.
Blood, 124(12), 1857-1867 (2015-01-22)
Platelet transfusions total >2.17 million apheresis-equivalent units per year in the United States and are derived entirely from human donors, despite clinically significant immunogenicity, associated risk of sepsis, and inventory shortages due to high demand and 5-day shelf life. To
Christian A Di Buduo et al.
Blood, 125(14), 2254-2264 (2015-01-13)
We present a programmable bioengineered 3-dimensional silk-based bone marrow niche tissue system that successfully mimics the physiology of human bone marrow environment allowing us to manufacture functional human platelets ex vivo. Using stem/progenitor cells, megakaryocyte function and platelet generation were
Volker F Achterberg et al.
The Journal of investigative dermatology, 134(7), 1862-1872 (2014-03-29)
Changes in the mechanical properties of dermis occur during skin aging or tissue remodeling and affect the activity of resident fibroblasts. With the aim to establish elastic culture substrates that reproduce the variable softness of dermis, we determined Young's elastic
Edward W Tekwa et al.
Lab on a chip, 15(18), 3723-3729 (2015-08-01)
Localized interactions are predicted to favour the evolution of cooperation amongst individuals within a population. One important factor that can localize interactions is habitat patchiness. We hypothesize that habitats with greater patchiness (greater edge-to-area ratio) can facilitate the maintenance of
Richard Karban et al.
The New phytologist, 204(2), 380-385 (2014-06-13)
Volatile communication between sagebrush (Artemisia tridentata) individuals has been found previously to reduce herbivory and to be more effective between individuals that are genetically identical or related relative to between strangers. The chemical nature of the cues involved in volatile

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service