Skip to Content
Merck
All Photos(2)

Key Documents

235709

Sigma-Aldrich

Octamethyltrisiloxane

98%

Synonym(s):

1,1,1,3,3,5,5,5-Octamethyltrisiloxane, Dimethylbis(trimethylsiloxy)silane, Pentamethyl(trimethylsiloxy)disiloxane, Pentamethyl(trimethylsilyloxy)disiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]2Si(CH3)2
CAS Number:
Molecular Weight:
236.53
Beilstein:
1753063
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.384 (lit.)

bp

153 °C (lit.)

mp

−82 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

InChI

1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3

InChI key

CXQXSVUQTKDNFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octamethyltrisiloxane, commonly referred to as octamethylcyclotetrasiloxane, falls under the category of siloxanes. Its versatile applications include serving as a solvent, coupling agent, and surface tension modifier in various industries. Furthermore, it plays a significant role in the production of adhesives, sealants, and lubricants.

Application

Octamethyltrisiloxane is used as a solvent for good dispersion of styrene and polydimethylsiloxane (PDMS), which are used in the fabrication of perovskite barium titanate inverse opal films.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - (External MSDS)

Flash Point(C)

29 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Aspromonte et al.
Analytical and bioanalytical chemistry, 412(11), 2537-2544 (2020-02-18)
Most of the reported methods for the analysis of volatile methylsiloxanes focus on their environmental fate or possible health effects, aiming at trace level analysis by using direct injection gas chromatography. However, system contamination as carry over and side reactions
Hunter Elliott et al.
Nature cell biology, 17(2), 137-147 (2015-01-27)
In many cases, cell function is intimately linked to cell shape control. We used endothelial cell branching morphogenesis as a model to understand the role of myosin II in shape control of invasive cells migrating in 3D collagen gels. We
Edyta Słupek et al.
Materials (Basel, Switzerland), 14(2) (2021-01-10)
The paper presents the screening of 20 deep eutectic solvents (DESs) composed of tetrapropylammonium bromide (TPABr) and glycols in various molar ratios, and 6 conventional solvents as absorbents for removal of siloxanes from model biogas stream. The screening was achieved
Gregory T Hohensee et al.
Nature communications, 6, 6578-6578 (2015-03-07)
The thermal conductance of interfaces between metals and diamond, which has a comparatively high Debye temperature, is often greater than can be accounted for by two-phonon processes. The high pressures achievable in a diamond anvil cell (DAC) can significantly extend
Chiara Ingrosso et al.
ACS applied materials & interfaces, 7(28), 15494-15505 (2015-07-08)
A novel UV-light-curable nanocomposite material formed of a methacrylic-siloxane resin loaded with 1 wt % oleic acid and 3-(trimethoxysilyl)propyl methacrylate silane (OLEA/MEMO)-coated TiO2 nanorods (NRs) has been manufactured as a potential self-curing structural coating material for protection of monuments and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service