Skip to Content
Merck
All Photos(1)

Key Documents

440191

Sigma-Aldrich

Hexamethyldisilazane

reagent grade, ≥99%

Synonym(s):

Bis(trimethylsilyl)amine, HMDS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiNHSi(CH3)3
CAS Number:
Molecular Weight:
161.39
Beilstein:
635752
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

125 °C (lit.)

SMILES string

C[Si](C)(C)N[Si](C)(C)C

InChI

1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3

InChI key

FFUAGWLWBBFQJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethyldisilane (HMDS) is a stable and effectivereagent for trimethylsilylation of hydrogen-labile substrates such as alcohols,amines, and carboxylic acids. It is commonly used for the protection of sensitivefunctional groups during chemical synthesis. Owing to its low cost, high vapor pressure, and safety, itis widely used as a precursor in chemical vapor deposition(CVD) reactions.

Application

Hexamethyldisilane (HMDS) can be used:
  • As a silylating agent in the trimethylsilylation of alcohols under nearly neutral reaction conditions.
  • To control the molecular weights of polypeptides during ring-opening polymerization of α-amino acid N-carboxyanhydrides.
  • To fabricate silicon carbonitride thin films by plasma-enhanced CVD.
  • For specimen preparation for scanning electron microscopy and the preparation of trimethylsilyl ethers from hydroxy compounds.

Features and Benefits

  • High chemicalstability and low molecular weight
  • Nontoxic andcost-effective reagent
  • Ammonia is theonly byproduct generated during silylation
  • Silylationreaction using HDMS is nearly neutral and does not need any precaution

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

52.5 °F - closed cup

Flash Point(C)

11.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and highly efficient trimethylsilylation of alcohols and phenols
with hexamethyldisilazane (HMDS) catalysed by in situ generated I2
using OxoneR /KI or cerium ammonium nitrate (CAN)/KI systems
under mild conditions
Eskandar K, et al.
Journal of Chemical Sciences (Bangalore), 123(5), 703-708 (2011)
F Braet et al.
Journal of microscopy, 186(Pt 1), 84-87 (1997-04-01)
Critical point drying (CPD) is a common method of drying biological specimens for scanning electron microscopy (SEM). Drying by evaporation of hexamethyldisilazane (HMDS) has been described as a good alternative. This method, however, is infrequently used. Therefore, we reassessed HMDS
Judith J de Vries et al.
International journal of molecular sciences, 21(19) (2020-10-01)
Subjects with diabetes mellitus (DM) have an increased risk of arterial thrombosis, to which changes in clot structure and mechanics may contribute. Another contributing factor might be an increased formation of neutrophil extracellular traps (NETs) in DM. NETs are mainly
Teresa Dean et al.
Biomaterials science, 8(11), 3078-3094 (2020-04-30)
Cancer associated fibroblasts (CAFs) are a major cellular component of the tumour stroma and have been shown to promote tumour cell invasion and disease progression. CAF-cancer cell interactions are bi-directional and occur via both soluble factor dependent and extracellular matrix
Clyde Savio Pinto et al.
Scientific reports, 9(1), 12191-12191 (2019-08-23)
Apical projections are integral functional units of epithelial cells. Microvilli and stereocilia are cylindrical apical projections that are formed of bundled actin. Microridges on the other hand, extend laterally, forming labyrinthine patterns on surfaces of various kinds of squamous epithelial

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the storage temperature of this reagent?

    1 answer
    1. This product may be stored at room temperature. For products with no specified storage temperature given on the label or product page, ambient storage may be assumed. Products that require controlled temperature storage, such as refrigeration or freezer conditions, storage temperatures are published on the product label, Certificate of Analysis, the Safety Data Sheet (SDS).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service