Skip to Content
Merck
All Photos(3)

Key Documents

456020

Sigma-Aldrich

Sodium formate

99.998% trace metals basis

Synonym(s):

Formic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein:
3595134
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.998% trace metals basis
form:
powder and chunks
Pricing and availability is not currently available.

Quality Level

Assay

99.998% trace metals basis

form

powder and chunks

impurities

≤25.0 ppm Trace Metal Analysis

mp

259-262 °C (lit.)

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Growth, Optical, Thermal, Mechanical and Electrical Properties of Anhydrous Sodium Formate Single Crystals: This study examines the properties of anhydrous sodium formate crystals for potential device applications, highlighting their thermal, optical, mechanical, and electrical characteristics (C Amuthambigai, CK Mahadevan, XS Shajan, 2016).
  • Hydroesterification of Alkenes with Sodium Formate and Alcohols Promoted by Cooperative Catalysis: Discusses a cooperative catalysis method using Ru3(CO)12 and 2-pyridinemethanol for converting alkenes into esters, employing sodium formate as a reagent (DS Kim, WJ Park, CH Lee, CH Jun, 2014).
  • Energy Conservation by Oxidation of Formate to Carbon Dioxide and Hydrogen via a Sodium Ion Current in a Hyperthermophilic Archaeon: Explores how sodium formate supports energy conservation in cellular processes by facilitating sodium ion currents in an archaeal model (JK Lim, F Mayer, SG Kang, 2014).
  • Eco-friendly, One-Pot Multicomponent Synthesis of Pyran Annulated Heterocyclic Scaffolds at Room Temperature Using Ammonium or Sodium Formate as Non-toxic Catalyst: Investigates the use of sodium formate as a non-toxic catalyst in the synthesis of pyran annulated heterocyclic compounds (G Brahmachari, S Laskar, 2014).
  • Four Compartment Mono Selective Electrodialysis for Separation of Sodium Formate from Industry Wastewater: Focuses on a novel method for recovering sodium formate from industrial effluents using electrodialysis, highlighting its effectiveness in environmental management (H Selvaraj, P Aravind, M Sundaram, 2018).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey Zakharov et al.
Kidney international, 86(1), 199-207 (2014-03-14)
During an outbreak of methanol poisonings in the Czech Republic in 2012, we were able to study methanol and formate elimination half-lives during intermittent hemodialysis (IHD) and continuous veno-venous hemodialysis/hemodiafiltration (CVVHD/HDF) and the relative impact of dialysate and blood flow
Nicolaas A Verwey et al.
Amyloid : the international journal of experimental and clinical investigation : the official journal of the International Society of Amyloidosis, 20(3), 179-187 (2013-07-09)
Abstract Amyloid β-peptide (Aβ) is a key molecule in Alzheimer's disease (AD). Reliable immunohistochemical (IHC) methods to detect Aβ and Aβ-associated factors (AAF) in brain specimens are needed to determine their role in AD pathophysiology. Formic acid (FA) pre-treatment, which
Therese Fagerqvist et al.
Journal of neurochemistry, 126(1), 131-144 (2013-02-01)
Inclusions of intraneuronal alpha-synuclein (α-synuclein) can be detected in brains of patients with Parkinson's disease and dementia with Lewy bodies. The aggregation of α-synuclein is a central feature of the disease pathogenesis. Among the different α-synuclein species, large oligomers/protofibrils have
Sai Duan et al.
Physical chemistry chemical physics : PCCP, 15(13), 4625-4633 (2013-02-21)
Local structures and adsorption energies of a formic acid molecule and its decomposed intermediates (H, O, OH, CO, HCOO, and COOH) on highly electrocatalytically active mushroom-like Au-core@Pd-shell@Pt-cluster nanoparticles with two atomic layers of the Pd shell and stoichiometric Pt coverage
An efficient CoAuPd/C catalyst for hydrogen generation from formic acid at room temperature.
Zhi-Li Wang et al.
Angewandte Chemie (International ed. in English), 52(16), 4406-4409 (2013-03-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service