Skip to Content
Merck
All Photos(1)

Key Documents

1.06443

Supelco

Sodium formate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium formate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHNaO2
CAS Number:
Molecular Weight:
68.01
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-488-0
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
550 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (perchloric acid titration)

form

solid

potency

>3000 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

loss

≤0.5% loss on drying, 150 °C

pH

7.0-8.5 (20 °C, 50 g/L in H2O)

mp

253 °C

solubility

550 g/L

density

1.92 g/cm3 at 20 °C

bulk density

635 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Ca: ≤0.0005%
Fe: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Analysis Note

Assay (perchloric acid titration): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water): 7.0 - 8.5
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Loss on drying (150 °C): ≤ 0.5 %
Corresponds to ACS, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service