Skip to Content
Merck
All Photos(2)

Documents

363081

Sigma-Aldrich

Poly(vinyl alcohol)

average Mw 85,000-124,000, 87-89% hydrolyzed

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

form

powder or crystals

mol wt

average Mw 85,000-124,000

falling ball

23-27 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyvinyl alcohol(PVA) is a linear hydrophilic synthetic polymer with a backbone composed of only carbon atoms. Due to its inertness and stability, it is considered safe and biocompatible. It is also biodegradable under both anaerobic and aerobic conditions. Owing to its unique properties such as adhesiveness, safety, strength, film-forming, swelling, and non-carcinogenicity, it is widely used in paper, adhesives,food, biomedical and pharmaceutical industries. The partially hydrolyzed PVA(87–89%) is more soluble in water and it has more flexibility and adhesion tohydrophobic surfaces.

Application

PVA has remarkable properties for a polymer to be used in drug delivery systems such as high surface stabilization, and low protein adsorption. The use of PVA in solid dispersion prevents crystal formation and improves the solubility of drugs. It can also be used as a granulating liquid to prevent slow crystal growth and nucleation. In conjunction with its copolymers, it can be useful for both hydrophilic and hydrophobic drugs. PVA hydrogels and microparticles can be used in drug delivery systems for cancer drugs.

PVA can be used as a surface stabilizer in preparing drug delivery carriers for the slow release of tetracycline.

Highly concentrated alginate/polyvinyl alcohol (PVA) bioinks can be used to prepare 3D bioprinting porous scaffolds. Partial diffusion of PVA sols from printed scaffolds creates a large number of micropores.

PVA can also be used as a water-soluble polymer template to synthesize nanosized polypyrrole via in situ polymerization. PVA generates stable dispersions of polypyrrole nanospheres in water and the size of nanospheres depends on the molecular weight and concentration of PVA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela Rivera-Hernández et al.
International journal of pharmaceutics, 600, 120478-120478 (2021-03-17)
Polyvinyl alcohol (PVA) is a biodegradable semicrystalline synthetic polymer that has been used for biomedical applications for several years. In the pharmaceutical area, PVA has been widely used to prepare solid dispersions to improve the solubility of drugs. Furthermore, it
Chihiro Fujiyabu et al.
Scientific reports, 9(1), 10653-10653 (2019-07-25)
Recent progress in whole genome sequencing has revealed that animals have various kinds of opsin genes for photoreception. Among them, most opsin genes have introns in their coding regions. However, it has been known for a long time that teleost
Adam C Wilkinson et al.
Nature, 571(7763), 117-121 (2019-05-31)
Multipotent self-renewing haematopoietic stem cells (HSCs) regenerate the adult blood system after transplantation1, which is a curative therapy for numerous diseases including immunodeficiencies and leukaemias2. Although substantial effort has been applied to identifying HSC maintenance factors through the characterization of
Gang Wang et al.
PloS one, 6(11), e27605-e27605 (2011-11-24)
Mucosal vaccination has been demonstrated to be an effective means of eliciting protective immunity against aerosol infections of foot and mouth disease virus (FMDV) and various approaches have been used to improve mucosal response to this pathogen. In this study
Piotr Hanczyc et al.
The journal of physical chemistry. B, 117(10), 2947-2954 (2013-02-15)
A stretched poly(vinyl alcohol) (PVA) film provides a unique matrix that enables also short DNA oligonucleotide duplex to be oriented and studied by linear dichroism (LD). This matrix further allows controlling DNA secondary structure by proper hydration (A or B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service