Skip to Content
Merck
All Photos(1)

Key Documents

437468

Sigma-Aldrich

Poly(ethylene glycol) dimethacrylate

average MN 750, cross-linking reagent polymerization reactions, methacrylate, 900-1100 ppm MEHQ as inhibitor

Synonym(s):

Polyethylene glycol, PEG dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor

form

liquid

Quality Level

mol wt

average Mn 750

contains

900-1100 ppm MEHQ as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.467

viscosity

67 cP(25 °C)(lit.)

bp

>200 °C/2 mmHg (lit.)

solubility

H2O: soluble

density

1.11 g/mL at 25 °C

Ω-end

methacrylate

α-end

methacrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

OCCO.CC(=C)C(O)=O

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adhesives, coatings, sealants, photoresists, solder masks and photopolymers.

Features and Benefits

Contributes flexibility.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pelagie M Favi et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1935-1944 (2013-03-19)
The culture of multipotent mesenchymal stem cells on natural biopolymers holds great promise for treatments of connective tissue disorders such as osteoarthritis. The safety and performance of such therapies relies on the systematic in vitro evaluation of the developed stem
C Aulin et al.
Laboratory animals, 47(1), 58-65 (2013-03-08)
Articular cartilage has a limited capacity for self-repair in adult humans, and methods used to stimulate regeneration often result in re-growth of fibrous cartilage, which has lower durability. No current treatment option can provide complete repair. The possibility of growth
Kwanghun Chung et al.
Nature methods, 10(6), 508-513 (2013-06-01)
With potential relevance for brain-mapping work, hydrogel-based structures can now be built from within biological tissue to allow subsequent removal of lipids without mechanical disassembly of the tissue. This process creates a tissue-hydrogel hybrid that is physically stable, that preserves
Enrique Barragan Garza et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 29(3), 166-172 (2013-03-01)
To conduct a feasibility study of the safety and efficacy of a corneal contouring inlay as a treatment for emmetropic presbyopia. The Raindrop corneal inlay (ReVision Optics, Inc., Lake Forest, CA) was implanted on the corneal stromal bed beneath a
Albert H Park et al.
The Laryngoscope, 123(4), 1043-1048 (2013-03-21)
To determine the resorption rate and biocompatibility characteristics of novel cross-linked hydrogel ventilation tubes and varied formulations of polyester ventilation tubes in a Chinchilla model. Animal Study. Three cross-linked glycosaminoglycan hydrogel ventilation tubes fabricated by cross-linking thiol-modified chondroitin sulfate or

Articles

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service