Skip to Content
Merck
All Photos(1)

Key Documents

202495

Sigma-Aldrich

Poly(ethylene glycol) methyl ether

average MN 750, methoxy, hydroxyl

Synonym(s):

Polyethylene glycol, Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene glycol) methyl ether, average Mn 750

vapor density

>1 (vs air)

Quality Level

vapor pressure

0.05 mmHg ( 20 °C)

form

paste
solid

mol wt

average Mn 750

refractive index

n20/D 1.459

viscosity

10.5 cSt(210 °F)(lit.)

transition temp

Tm 30 °C

density

1.094 g/mL at 25 °C

Ω-end

hydroxyl

α-end

methoxy

SMILES string

O(CCO)C

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) methyl ether can be used:
  • As a chain transfer agent to synthesize amphiphilic block copolymers by metal-free ring-opening oligomerization.
  • As a precursor to prepare retinoic acid-polyethylene glycol nanoassembly as an efficient drug delivery system.
  • To prepare diblock copolymer with polylactic acid, which can be applied in the field of tissue engineering and drug delivery.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Liu et al.
International journal of pharmaceutics, 443(1-2), 175-182 (2013-01-05)
This work aims to develop curcumin (Cur) loaded biodegradable self-assembled polymeric micelles (Cur-M) to overcome poor water solubility of Cur and to meet the requirement of intravenous administration. Cur-M were prepared by solid dispersion method, which was simple and easy
Yi Wei et al.
Langmuir : the ACS journal of surfaces and colloids, 28(39), 13984-13992 (2012-09-04)
The microcosmic mechanisms of protein (recombinant human growth hormone, rhGH) incomplete release and stability from amphiphilic poly(monomethoxypolyethylene glycol-co-D,L-lactide) (mPEG-PLA, PELA) microspheres were investigated. PELA with different hydrophilicities (PELA-1, PELA-2, and PELA-3) based on various ratios of mPEG to PLA were
Yiyi Yu et al.
Journal of pharmaceutical sciences, 102(3), 1054-1062 (2013-01-03)
To promote the application of methoxy poly(ethylene glycol)-cholesterol (mPEG-Chol), mPEG-Chol was used to prepare core-shell micelles encapsulating poorly water-soluble docetaxel (DTX-PM) by modified cosolvent evaporation method. Approaches to enhance DTX entrapment efficiency (EE) and minimize particle size were investigated in
Pengxiang Zhao et al.
Chemical communications (Cambridge, England), 49(31), 3218-3220 (2013-03-14)
"Click" chemistry now offers access to a great variety of triazoles, and the first example of a strategy to stabilize gold nanoparticles (AuNPs) with a new 1,2,3-triazole-mPEG ligand is developed here together with preliminary examples of possible applications.
Lina Du et al.
Anti-cancer drugs, 24(2), 172-180 (2012-09-20)
A functionalized poly(amidoamine) (PAMAM) nanocarrier was designed and prepared to deliver anticancer drugs. The nanocarrier is a copolymer with a core-shell structure with 3.0 G PAMAM as the core and sequentially conjugated poly(2-(N,N-diethylamino)ethyl methacrylate) (pDEA) and methoxy-poly(ethylene glycol) 2000 (mPEG)

Articles

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

See All

Questions

  1. What is the molecular weight in g/mol for Poly(ethylene glycol) methyl ether, Mn = 750?

    1 answer
    1. Yes, as listed in the PROPERTIES section, under 'mol wt', the number average molecular weight is 750.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service