Skip to Content
Merck
All Photos(1)

Key Documents

569615

Sigma-Aldrich

(3-Chloropropyl)triethoxysilane

95%

Synonym(s):

(gamma-Chloropropyl)triethoxysilane, 3-(Triethoxysilyl)-1-chloropropane, 3-(Triethoxysilyl)propyl chloride, 3-Chloropropropyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CH2Si(OCH2CH3)3
CAS Number:
Molecular Weight:
240.80
Beilstein:
1754992
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.418 (lit.)

density

1.000 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CCO[Si](CCCCl)(OCC)OCC

InChI

1S/C9H21ClO3Si/c1-4-11-14(12-5-2,13-6-3)9-7-8-10/h4-9H2,1-3H3

InChI key

KSCAZPYHLGGNPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

98.6 °F

Flash Point(C)

37 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minghua Lu
The Analyst, 142(18), 3477-3483 (2017-08-23)
An electrochemical sensing platform was designed to monitor telomerase activity in HeLa cells, using bioresponsively controlled cargo release from magnetic mesoporous silica nanocontainers (MMSNs). The aminated MMSNs were first synthesized by a wet-chemistry method, then methylene blue (indicator) molecules were
Nicholas G Fischer et al.
Coatings (Basel, Switzerland), 10(6) (2020-08-29)
Many chemical routes have been proposed to immobilize peptides on biomedical device surfaces, and in particular, on dental implants to prevent peri-implantitis. While a number of factors affect peptide immobilization quality, an easily controllable factor is the chemistry used to
Lin Zhou et al.
Colloids and surfaces. B, Biointerfaces, 160, 581-588 (2017-10-14)
The inflammatory reaction around the implant after implant placement is important not only for osseointegration but also for long-term implant survivals. In our study, GL13K, an antimicrobial peptide, was immobilized onto titanium surfaces to improve its anti-inflammatory properties. The method
Kheibar Dashtian et al.
Journal of colloid and interface science, 494, 114-123 (2017-02-06)
A selective adsorbent based on the modification of mesoprous SBA-15 with N,N'-bis(salicylidene)-1,3-ethylenediamine Schiff base and decorated with Fe
Younes Ellahioui et al.
Dalton transactions (Cambridge, England : 2003), 48(18), 5940-5951 (2018-09-14)
A series of nanomaterials based on mesoporous silica have been synthesised and functionalised with a photoactive polypyridyl ruthenium(ii) complex, namely [Ru(bipy)2-dppz-7-hydroxymethyl][PF6]2 (bipy = 2,2'-bipyridine, dppz = dipyrido[3,2-a:2',3'-c]phenazine), by various methods. The functionalisation reactions were based on the covalent binding to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service