Skip to Content
Merck
All Photos(1)

Documents

385514

Sigma-Aldrich

(S)-(−)-2-Bromopropionic acid

99% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(Br)CO2H
CAS Number:
Molecular Weight:
152.97
Beilstein:
1720261
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −25°, neat

refractive index

n20/D 1.470

bp

78 °C/4 mmHg (lit.)

mp

−35 °C (lit.)

density

1.696 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](Br)C(O)=O

InChI

1S/C3H5BrO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/t2-/m0/s1

InChI key

MONMFXREYOKQTI-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for the direct enantiomeric assay of carboxylic acids by proton NMR.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron Asymmetry, 3, 25-25 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service