Skip to Content
Merck
All Photos(1)

Documents

306800

Sigma-Aldrich

(R)-(+)-2-Chloropropionic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(Cl)CO2H
CAS Number:
Molecular Weight:
108.52
Beilstein:
1720258
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

optical activity

[α]20/D +14°, neat

refractive index

n20/D 1.4345 (lit.)

bp

185-188 °C (lit.)

density

1.258 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](Cl)C(O)=O

InChI

1S/C3H5ClO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/t2-/m1/s1

InChI key

GAWAYYRQGQZKCR-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-2-Chloropropionic acid can be used as a starting material in the synthesis of:       
  • Thiolactic acid, a key intermediate, which is used for the preparation of 2(R),5(S)-oxathiolanones by reacting with pivalaldehyde.       
  • (R)-2-chloropropionyl chloride, a key intermediate, which is utilized in the synthesis of (R)-4,5-dihydro-5-methylpyridazin-3(2H)-one derivative with pyrazolopyridine ring as a potential phosphodiesterase inhibitor.       
  • 2-Chloropropionamide derivatives as protein disulfide isomerase (PID) inhibitors.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dharmaraja Allimuthu et al.
ACS chemical biology, 12(8), 2124-2131 (2017-06-15)
Resurgent interest in covalent target engagement in drug discovery has demonstrated that small molecules containing weakly reactive electrophiles can be safe and effective therapies. Several recently FDA-approved drugs feature an acrylamide functionality to selectively engage cysteine side chains of kinases
Jill M McFadden et al.
Organic letters, 4(22), 3859-3862 (2003-02-26)
[formula: see text] A new and efficient asymmetric synthesis of naturally occurring (5R)-thiolactomycin (1) using D-alanine as the source of chirality is described.
A short and efficient synthesis of a chiral pyridazinone derivative by the chiral-pool method
Yoshida N, et al.
Synthesis, 2004(10), 1554-1556 (2004)
Eugénie Grigorian et al.
Frontiers in microbiology, 12, 725997-725997 (2021-10-09)
L-2-halocid dehalogenases (L-2-HADs) have been mainly characterized from terrestrial polluted environments. By contrast, knowledge is still scarce about their role in detoxification of predominant halocarbons in marine environments. Here, phylogenetic analyses showed a wide diversity of homologous L-2-HADs, especially among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service