Skip to Content
Merck
All Photos(1)

Documents

370886

Sigma-Aldrich

1,1,1,3,5,5,5-Heptamethyltrisiloxane

97%

Synonym(s):

2,2,4,6,6-Pentamethyl-3,5-dioxa-2,4,6-trisilaheptane, 2-Hydro-1,1,1,2,3,3,3-heptamethyltrisiloxane, Bis(trimethylsiloxy)methylsilane, Heptamethylhydrotrisiloxane, Methylbis(trimethylsiloxy)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]2SiHCH3
CAS Number:
Molecular Weight:
222.50
Beilstein:
1748455
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.382 (lit.)

bp

142 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

SMILES string

C[SiH](O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C7H22O2Si3/c1-10(8-11(2,3)4)9-12(5,6)7/h10H,1-7H3

InChI key

QNWOFLWXQGHSRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,1,1,3,5,5,5-Heptamethyltrisiloxane can be used for the aromatic C-H silylation of arenes in the presence of a platinum complex catalyst.
It can also be used as a reagent to synthesize:
  • 3-Octyl-1,1,1,3,5,5,5-heptamethyltrisiloxane, an agricultural adjuvant and sensory performance enhancer used in cosmetic formulations.
  • Isoindigo-based conjugated polymer with siloxane-terminated solubilizing group.
  • Monosiloxy esters with a variety of acids.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient Pd-catalyzed route to silyl esters
Chauhan M, et al.
Organic Letters, 2(8), 1027-1029 (2000)
Platinum-catalyzed aromatic C-H silylation of arenes with 1, 1, 1, 3, 5, 5, 5-Heptamethyltrisiloxane
Murata M, et al.
Chemistry Letters (Jpn), 36(7), 910-911 (2007)
Siloxane-terminated solubilizing side chains: bringing conjugated polymer backbones closer and boosting hole mobilities in thin-film transistors
Mei J, et al.
Journal of the American Chemical Society, 133(50), 20130-20133 (2011)
Iron catalysts for selective anti-Markovnikov alkene hydrosilylation using tertiary silanes
Tondreau AM, et al.
Science (New York, N.Y.), 335(6068), 567-570 (2012)
Kairui Liu et al.
Nature communications, 10(1), 996-996 (2019-03-03)
While numerous single atoms stabilized by support surfaces have been reported, the synthesis of in-situ reduced discrete metal atoms weakly coordinated and stabilized in liquid media is a more challenging goal. We report the genesis of mononuclear electron deficient Pt1(0)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service