Skip to Content
Merck
All Photos(1)

Key Documents

328650

Sigma-Aldrich

Potassium tert-butoxide solution

1.0 M in THF

Synonym(s):

Potassium tert-butylate, Potassium-2-methylpropan-2-olate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COK
CAS Number:
Molecular Weight:
112.21
Beilstein:
3556712
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

reaction suitability

core: potassium

concentration

1.0 M in THF

density

0.902 g/mL at 25 °C

SMILES string

[K+].CC(C)(C)[O-]

InChI

1S/C4H9O.K/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LPNYRYFBWFDTMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium tert-butoxide solution is a strong non nucleophilic base. It is capable of deprotonating carbon and other Bronsted acids. Reagent solutions in tert-BuOh may be prepared by reacting anhydrous alcohol with potassium in nitrogen atmosphere.

Application

Potassium tert-butoxide solution(t-BuOK) can be used:
  • As a catalyst for the interesterification of rapeseed oil with methyl acetate.
  • To promote Sommelet–Hauser rearrangements of N-benzylic amino acid-derived ammonium ylides under mild conditions.
  • As a metal-organic precursor in the synthesis of KNbO3 ferroelectric films by the chemical vapor deposition method.
  • To prepare 3-potassiooxamethylpyridine catalyst.
  • As a strong alkoxide base reagent.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step methodology for the synthesis of FA picolinyl esters from intact lipids.
Destaillats F and Angers P
Journal of the American Oil Chemists' Society, 79(3), 253-256 (2002)
Eiji Tayama et al.
Organic & biomolecular chemistry, 6(20), 3673-3680 (2008-10-10)
The Stevens rearrangement of N-allylic alpha-aryl amino acid-derived ammonium salts and the Sommelet-Hauser rearrangement of N-benzylic alpha-alkyl amino acid-derived ammonium salts are shown to proceed with remarkably high levels of diastereoselectivity. The methods presented in this work provide new routes
Wen-Sen He et al.
Bioresource technology, 114, 1-5 (2012-04-03)
The effects of solvents with different log P values, and of lipases on the synthesis of water-soluble plant stanol derivatives were investigated. Results showed that conversion in solvents with log P<0.37 was mainly controlled by the hydrophobicity of the solvent
Brian C Anderson et al.
Biomaterials, 23(22), 4345-4352 (2002-09-11)
Several homopolymers and copolymers of 2-(diethylamino)ethyl methacrylate (DEAEM) and poly(ethylene glycol) methyl ether methacrylate (PEGMEM) were synthesized using anionic polymerization initiated by potassium t-butoxide. The polymers were characterized by average molecular weight, polydispersity and monomeric unit composition. A very narrow
Astrid Beyer et al.
Organic letters, 14(15), 3948-3951 (2012-07-17)
Potassium tert-butoxide-mediated intramolecular α-arylations of fluoro- and chloro-substituted anilides provide oxindoles in DMF at 80 °C. In this manner, diversely substituted products have been obtained in moderate to high yields.

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service