Skip to Content
Merck
All Photos(1)

Key Documents

659878

Sigma-Aldrich

Potassium tert-butoxide

sublimed grade, 99.99% trace metals basis

Synonym(s):

Potassium tert-butylate, Potassium t-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COK
CAS Number:
Molecular Weight:
112.21
Beilstein:
3556712
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Quality Level

vapor pressure

1 mmHg ( 220 °C)

Assay

99.99% trace metals basis

form

solid

reaction suitability

core: potassium

mp

256-258 °C (dec.) (lit.)

SMILES string

[K+].CC(C)(C)[O-]

InChI

1S/C4H9O.K/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LPNYRYFBWFDTMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium tert-butoxide is a strong alkoxide base, it can deprotonate carbon and other Brφnsted acids. It is a relatively poor nucleophile.
Total impurities: may contain up to 5,000 ppm Sodium

Application

Mizoroki-Heck-Type reactions are mediated by potassium tert-butoxide. potassium tert-butoxide can commence the anionic polymerization of carbazolyl-substituted oxiranes.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anionic polymerization of carbazolyl-substituted oxiranes initiated by potassium alkalide, potassium tert-butoxide and potassium hydride.
Buika G, et al.
Macromolecular Chemistry and Physics, 196(4), 1287-1293 (1995)
Einav Amit et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(57), 13046-13052 (2020-04-29)
N-heterocyclic carbenes (NHCs) have emerged as a unique molecular platform for the formation of self-assembled monolayers (SAMs) on various surfaces. However, active carbene formation requires deprotonation of imidazolium salt precursors, which is mostly facilitated by exposure of the salt to
Eiji Tayama et al.
Organic & biomolecular chemistry, 6(20), 3673-3680 (2008-10-10)
The Stevens rearrangement of N-allylic alpha-aryl amino acid-derived ammonium salts and the Sommelet-Hauser rearrangement of N-benzylic alpha-alkyl amino acid-derived ammonium salts are shown to proceed with remarkably high levels of diastereoselectivity. The methods presented in this work provide new routes
Wen-Sen He et al.
Bioresource technology, 114, 1-5 (2012-04-03)
The effects of solvents with different log P values, and of lipases on the synthesis of water-soluble plant stanol derivatives were investigated. Results showed that conversion in solvents with log P<0.37 was mainly controlled by the hydrophobicity of the solvent
Astrid Beyer et al.
Organic letters, 14(15), 3948-3951 (2012-07-17)
Potassium tert-butoxide-mediated intramolecular α-arylations of fluoro- and chloro-substituted anilides provide oxindoles in DMF at 80 °C. In this manner, diversely substituted products have been obtained in moderate to high yields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service